Environmental Express® Color-Coded BOD Stoppers
Easily identify what's in the BOD bottle based on the color of the stopper
  • Pack of 25 stoppers
These plastic stoppers work with all brands of glass and plastic, 60 mL and 300 mL BOD bottles. The colored cap on the BOD stopper can be used to differentiate your samples.
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
BOD Bottle Stopper, Plastic, Blue Cap, 25/PK
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)
COMPARE
BOD Bottle Stopper, Plastic, Green Cap, 25/PK
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)
COMPARE
BOD Bottle Stopper, Plastic, Red Cap, 25/PK
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)