Environmental Express D1025-BLUE BOD Bottle Stopper, Plastic, Blue Cap, 25/PK
Representative image only.
Environmental Express D1025-BLUE BOD Bottle Stopper, Plastic, Blue Cap, 25/PK
Environmental Express | Item: 5320036
Easily identify what's in the BOD bottle based on the color of the stopper
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)
£98.88GBP / Pkg of 25
£118.66GBP (incl VAT)
Product Description
Key Features
  • Pack of 25 stoppers
  • Model number D1025-BLUE
More About this Item
These plastic stoppers work with all brands of glass and plastic, 60 mL and 300 mL BOD bottles. The colored cap on the BOD stopper can be used to differentiate your samples.
Specifications
  • ColorBlue
  • MaterialPlastic
  • DescriptionBOD Bottle Stopper, Plastic, Blue Cap, 25/PK