Cole-Parmer® Flexicon Style 316L Stainless Steel Filling Needles
Engineered for high precision and accuracy in filling applications
  • Ensure accurate dosing while minimizing waste
  • Provides 100% coverage across the fill-finish arena
  • Made from premium 316L stainless steel with polycarbonate housing
  • 70% IPA cleaned in ISO Class 7 cleanroom prior to double bagging
  • USP Class VI, FDA 21 CFR177.1580, REACH, and RoHS compliant
  • Latex and phthalate free
The single-use filling needles come ready to use—simply open, sterilize, and place in filling machines. Needles go through meticulous quality control checks to ensure consistent, repeatable performance across batches. Needles are constructed of high-quality 316L stainless steel needles with polycarbonate housing in an oil- and adhesive-free manufacturing process, minimizing introduction of foreign particles into the filling environment. Needle straightness of <2 mm/M allows for high-speed filling.
£304.45GBP / Each
£365.34GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
For Tube Inside Diameter (ID) (mm) Inside Diameter (ID) (mm) Outside Diameter (OD) (mm)
Availability Pricing
COMPARE
0.8 0.6 1
£304.45GBP / Each
£365.34GBP (incl VAT)
COMPARE
1.2 1 1.5
£304.45GBP / Each
£365.34GBP (incl VAT)
COMPARE
1.6 1.6 2.1
£304.45GBP / Each
£365.34GBP (incl VAT)
COMPARE
3.2 3 3.8
£304.45GBP / Each
£365.34GBP (incl VAT)
COMPARE
4.8 4.5 5.5
£304.45GBP / Each
£365.34GBP (incl VAT)
COMPARE
6.4 6 7
£304.45GBP / Each
£365.34GBP (incl VAT)