Cole-Parmer® Cooling Blocks
Quit running to the freezer for more ice
  • Cool or heat your samples quickly—blocks adjust to desired temperature within minutes
  • Eliminate uneven cooling
  • Keep samples upright and organized while controlling temperature
  • Contoured wells hold tubes and vials snugly for excellent temperature convection
Place blocks on ice, dry ice, liquid nitrogen, or in a water bath. These anodized aluminum blocks can keep samples at or near 1°C for up to 8 hours in properly packed ice, or for 3 to 7 hours in dry ice. Choose the blocks that meet your application needs — configurations are available for 1.5-/2.0-mL microcentrifuge tubes and cryogenic vials, 0.5-mL tubes, and 96-well or 364-well plates.
£115.62 - £173.75GBP / Each
£138.74 - £208.50GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size Description
Availability Pricing
COMPARE
15 1.5 to 2.0 mL Microcentrifuge Aluminum Cooling Block, 15-Well for 1.5/2.0 mL Tubes
£124.02GBP / Each
£148.82GBP (incl VAT)
COMPARE
15 1.5 to 2.0 mL Cryogenic Vials Aluminum Cooling Block, 15-Well for Cryogenic Vials
£115.62GBP / Each
£138.74GBP (incl VAT)
COMPARE
24 0.5 mL Aluminum Cooling Block, 24-Well for 0.5 mL Tubes
£149.35GBP / Each
£179.22GBP (incl VAT)
COMPARE
96 0.2 mL Aluminum Cooling Block, 96-Well for 0.2 mL Tubes
£143.96GBP / Each
£172.75GBP (incl VAT)
COMPARE
384 PCR Plates Aluminum Cooling Block, 384-Well PCR Plate
£173.75GBP / Each
£208.50GBP (incl VAT)