Cole-Parmer Aluminum Cooling Block, 15-Well for Cryogenic Vials
Representative image only.
Cole-Parmer Aluminum Cooling Block, 15-Well for Cryogenic Vials
Cole-Parmer | Item: 6361502
Quit running to the freezer for more ice
£115.62GBP / Each
£138.74GBP (incl VAT)
£115.62GBP / Each
£138.74GBP (incl VAT)
Buy It Together
Current Item
Cole-Parmer Aluminum Cooling Block, 15-Well for Cryogenic Vials
Cole-Parmer Aluminum Cooling Block, 15-Well for Cryogenic Vials
6361502
£115.62GBP / Each
£138.74GBP (incl VAT)
+
Cole-Parmer Transport Box, 5 L
0372604
£314.85GBP / Each
£377.82GBP (incl VAT)
+ + =
Total Price 430.47 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Cool or heat your samples quickly—blocks adjust to desired temperature within minutes
  • Eliminate uneven cooling
  • Keep samples upright and organized while controlling temperature
  • Contoured wells hold tubes and vials snugly for excellent temperature convection
More About this Item
Place blocks on ice, dry ice, liquid nitrogen, or in a water bath. These anodized aluminum blocks can keep samples at or near 1°C for up to 8 hours in properly packed ice, or for 3 to 7 hours in dry ice. Choose the blocks that meet your application needs — configurations are available for 1.5-/2.0-mL microcentrifuge tubes and cryogenic vials, 0.5-mL tubes, and 96-well or 364-well plates.
Specifications
  • Tubes Held15
  • Tube Size1.5 to 2.0 mL Cryogenic Vials
  • ColorBlue
  • DescriptionAluminum Cooling Block, 15-Well for Cryogenic Vials