Trajan SGE EVOL MEPS Electronic Syringe; 50 µL
Representative image only.
Trajan SGE EVOL MEPS Electronic Syringe; 50 µL
Trajan | Mfr: 2910027| Item: 1794008
A faster method for sample separation
Call For Price
Call For Price
Buy It Together
Current Item
Trajan SGE EVOL MEPS Electronic Syringe; 50 µL
Trajan SGE EVOL MEPS Electronic Syringe; 50 µL
1794008
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Use less solvent and reduce waste with microliter needles
  • Smaller volumes process quicker
  • Semiautomated functions minimize operator intervention
  • Volume: 50 µL
More About this Item
This handheld digital syringe features a familiar touch wheel user interface and includes a full-color screen. Use with 5 µL to 1 mL MEPS syringes. Easily and quickly change XCHANGE® enabled syringes. Custom programming function allows process to be validated and semiautomated before transition to a fully automated platform. Extracted samples are compatible with gas and liquid chromatography systems.
Available for sale in the Middle East, Europe, and United States only
Specifications
  • Syringe Capacity (µL)50
  • Needle IncludedNo
  • SterileNo
  • DescriptionMEPS Electronic Syringe; 50 µL