Trajan SGE EVOL MEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip
Representative image only.
Trajan SGE EVOL MEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip
Trajan | Mfr: 2910028| Item: 1794009
A faster method for sample separation
Call For Price
Call For Price
Buy It Together
Current Item
Trajan SGE EVOL MEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip
Trajan SGE EVOL MEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip
1794009
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Use less solvent and reduce waste with microliter needles
  • Smaller volumes process quicker
  • Semiautomated functions minimize operator intervention
  • Volume: 100 µL
More About this Item
This handheld digital syringe features a familiar touch wheel user interface and includes a full-color screen. Use with 5 µL to 1 mL MEPS syringes. Easily and quickly change XCHANGE® enabled syringes. Custom programming function allows process to be validated and semiautomated before transition to a fully automated platform. Extracted samples are compatible with gas and liquid chromatography systems.
Available for sale in the Middle East, Europe, and United States only
Specifications
  • Syringe Capacity (µL)100
  • Needle Tip StyleBevel
  • Needle IncludedYes
  • Needle Gauge25
  • SterileNo
  • DescriptionMEPS Electronic Syringe, 100 µL, Removable Needle, 25G; Bevel Tip