Thermo Scientific 9606BNWP Sure-Flow Ion-Selective Electrode; Cyanide
Representative image only.
Thermo Scientific 9606BNWP Sure-Flow Ion-Selective Electrode; Cyanide
Thermo Scientific | Mfr: 9606BNWP| Item: 0571309
Orion-designed for optimum performance and the most comprehensive ISE selection
£1,106.32GBP / Each
£1,327.58GBP (incl VAT)
£1,106.32GBP / Each
£1,327.58GBP (incl VAT)
Buy It Together
Current Item
Thermo Scientific 9606BNWP Sure-Flow Ion-Selective Electrode; Cyanide
Thermo Scientific 9606BNWP Sure-Flow Ion-Selective Electrode; Cyanide
0571309
£1,106.32GBP / Each
£1,327.58GBP (incl VAT)
+
Thermo Scientific 900062 Optimum Results B ISE Fill Solution; Cl/CN/Pb/ Ag/S
0571332
£161.87GBP / Each
£194.24GBP (incl VAT)
+
Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride
0572111
£1,081.05GBP / Each
£1,297.26GBP (incl VAT)
+
Cole-Parmer Replaceable Membrane ISE Probe; Ammonium
2750500
£407.27GBP / Each
£488.72GBP (incl VAT)
+
Thermo Scientific Dual Star pH/ISE/mV Meter Only
5882500
£2,220.47GBP / Each
£2,664.56GBP (incl VAT)
+
Thermo Scientific STAR A214 Star A214 pH/ISE Benchtop Meter Only
5882526
£1,758.78GBP / Each
£2,110.54GBP (incl VAT)
=
Total Price 6735.76 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • ionplus™ Sure-Flow™ Combination Ion-Selective Electrodes eliminate fumbling and tangles from multiple cables
  • Combination Ion-Selective Electrodes feature both half-cell and reference cell in one electrode
  • Half-Cell Ion-Selective Electrodes are the classic means of taking ion measurements
  • 1 year warranty
More About this Item
Combination Ion-Selective Electrodes are easier to use than reference and half-cells—eliminating fumbling and tangles from multiple cables. Sure-flow® technology provides stable, reproducible results. The easy-to-clean design reduces the risk of inaccurate readings due to clogging of the reference junction. All electrodes include a BNC connector, a 3-ft cable, and manual. Complete systems require-the purchase of a standardizing solution, filling solution for the reference electrode, ionic strength adjustor, and pH adjusters. Chloride, cyanide, Sulfide, nitrate, and fluoride are approved for EPA methods for analysis of wastewater and for NPDES permitting.
1 Year Warranty
Specifications
  • Electrode TypeSolid-state
  • Reference JunctionSure-flow
  • Reference ElectrolyteOptimum filling solution B
  • Ion-Selective ElectrodeCyanide
  • Min pH (pH)13
  • Max Temperature (° C)50
  • ConnectorBNC (waterproof when used with Thermo Scientific Orion Star Series meters)
  • DescriptionSure-Flow Ion-Selective Electrode; Cyanide