Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L

Representative image only.

Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L

Reagents | Mfr: C2346500-1A| Item: 6228626

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price

Buy It Together

Current Item

Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L
Reagents Deionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L
6228626

Call For Price
+ + =
Total Price 0 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Water meets ASTM Type I D 1193 reagent grade certification
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • 1 L poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaH2O
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDihydrogen oxide, Dihydrogen monoxide
  • Volume (mL)1000
  • Vapor Pressure (torr)23.8
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolAmbient
  • CAS Number7732-18-5
  • Shelf Life (days)540
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • Refractive Index1.333
  • DescriptionDeionized Water, ACS Reagent, ASTM Type I (D 1193), Poly Bottle, 1 L
Documents & Certificates
Reviews
Q&A