NSI Lab Solutions QCI-034B Trace Metals Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCI-034B Trace Metals Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120364

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • Supplied in duplicate for convenience and value
  • Two 21-mL concentrate set for analysis trace metals
  • Each ampule produces 2 L of sample

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

For determination of the following elements:
Aluminum: 200 to 4000 µg/L
Antimony: 90 to 900 µg/L
Arsenic: 90 to 900 µg/L
Barium: 100 to 2500 µg/L
Beryllium: 50 to 500 µg/L
Boron: 800 to 2000 µg/L
Cadmium: 100 to 1000 µg/L
Chromium: 100 to 1000 µg/L
Cobalt: 100 to 1000 µg/L
Copper: 100 to 1000 µg/L
Iron: 200 to 4000 µg/L
Lead: 100 to 1500 µg/L
Lithium: 50 to 500 µg/L
Manganese: 200 to 2000 µg/L
Molybdenum: 60 to 600 µg/L
Nickel: 200 to 2000 µg/L
Selenium: 100 to 1000 µg/L
Silver: 100 to 1000 µg/L
Strontium: 50 to 500 µg/L
Thallium: 80 to 800 µg/L
Tin: 200 to 2000 µg/L
Titanium: 60 to 300 µg/L
Vanadium: 50 to 2000 µg/L
Zinc: 300 to 2000 µg/L

Specifications
  • Element/IonAluminum, Antimony, Arsenic, Barium, Beryllium, Boron, Cadmium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Manganese, Molybdenum, Nickel, Selenium, Silver, Strontium, Thallium, Tin, Titanium, Vanadium, Zinc
  • FormLiquid
  • Volume (mL)21
  • Packaging2/PK
  • Storage SymbolStore between 15 and 30 °C
  • UN Number2031
  • Shelf Life (months)24
  • DescriptionTrace Metals Wastewater PT Express™; 2/PK
Reviews
Q&A