NSI Lab Solutions 10K-0012 E. coli NCTC 12933 Pharmaceutical Culture, 10,000 CFU; 10/Pk

Representative image only.

NSI Lab Solutions 10K-0012 E. coli NCTC 12933 Pharmaceutical Culture, 10,000 CFU; 10/Pk

NSI Lab Solutions | Item: 0500635

A single pellet is formulated to recover approximately 10,000 CFU

Call For Price

Call For Price
Product Description

Includes

Ten vials with 1 pellet per vial.

Key Features

  • Reliable and easy to use – 10,000 CFU per pellet
  • Hydration fluid dissolves culture pellet in less than a minute
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • Equivalent to WDCM 0012 and ATCC 8739

More About this Item

These microbiological reference materials are designed for use as quantitative control for pharmaceutical culture analysis. The single strain ISO 17034 certified reference material is formulated to recover approximately 10,000 CFU/pellet by cultural plating techniques. For your convenience, each pack includes ten culture pellets individually vacuum sealed in 5 mL glass vials. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Rehydration of a single 10,000 CFU/pellet in 100 mL fluid would provide 100 CFU per mL. Following this prescribed rehydration scheme provides up to 100 replicate tests per hydrated pellet. Extra activity listed on Certificate of Analysis (CoA). Store in the freezer.

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

ATCC is a registered trademark of American Type Culture Collection.

NCTC and NCPF reference cultures obtained from the National Collection of Type Cultures and National Collection of Pathogenic Fungi, two of the culture collections of the UK Health Security Agency.

WDCM reference cultures obtained from the WFCC-MIRCEN World Data Centre for Organisms.

Specifications
  • OrganismE. coli
  • Organism typeNCTC 12933, WDCM 0012, ATCC 8739
  • Packaging10/Pk
  • Product TypePharmaceutical Cultures
  • Shelf Life Months (months)24
  • Shipping requirementFrozen ice packs
  • Storage requirementStore between –20 and –10°C
  • DescriptionE. coli NCTC 12933 Pharmaceutical Culture, 10,000 CFU; 10/Pk
Reviews
Q&A