Analytik Jena Pen-Ray 98000801 Lamp shield, 0.04" ID opening 0.04" (0.1 cm) ID opening
Representative image only.
Analytik Jena Pen-Ray 98000801 Lamp shield, 0.04" ID opening 0.04" (0.1 cm) ID opening
Analytik Jena | Mfr: 98-0008-01| Item: 9760650
£70.77GBP / Each
£84.92GBP (incl VAT)
£70.77GBP / Each
£84.92GBP (incl VAT)
Buy It Together
Current Item
Analytik Jena Pen-Ray 98000801 Lamp shield, 0.04" ID opening 0.04" (0.1 cm) ID opening
Analytik Jena Pen-Ray 98000801 Lamp shield, 0.04" ID opening 0.04" (0.1 cm) ID opening
9760650
£70.77GBP / Each
£84.92GBP (incl VAT)
+
Analytik Jena Pen-Ray 90001201 Standard UV lamp; 254 nm, 2.13"
9760600
£458.54GBP / Each
£550.25GBP (incl VAT)
+ + +
Cole-Parmer Handheld UV Lamp, 6 Watt, 254/365 nm, AC Operated; 110 VAC, 50/60 Hz
0981807
£331.86GBP / Each
£398.23GBP (incl VAT)
+ + + =
Total Price 861.17 ADD 8 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Keep your ultraviolet equipment running and dependable with replacement bulbs and accessories.
This accessory is available for deluxe model 97606-00.
Use to direct UV light through a point-like aperture or small window opening. Shield also protects tube from breakage.
Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.
1 Year Warranty
Specifications
  • Compatible With97606-00
  • Diameter (InchShort)3/64
  • Diameter (mm)10
  • DescriptionLamp shield, 0.04" ID opening 0.04" (0.1 cm) ID opening