Weighing Unit Conversion Table
Weighing Unit Conversion Table
To convert from grams to: Abbreviation multiply by
carats ct 5.0
dram — 0.564383
grains gn 15.432358
momme mom 0.2666666
ounces oz 0.0352739
pennyweights dwt 0.6430149
pounds lb 0.0022046
scruple — 0.7716179
tael (Hong Kong) tlh 0.0264554
tola (India) tol 0.0857353
tael (Singapore) tls 0.0264599
tael (Taiwan) tlt 0.0266666
troy ounces oz t 0.0321507
troy pounds lb t 0.0026792