Thread to barbed adapters for sterile applications
Adapter types for your soft-wall and flexible tubing
Connect a variety of softwall tubing sizes to threaded ports with semi-rigid tubing
  • Available from 1.0mm ID to 8.0mm ID
  • Available with 1/4"-28 flat bottom, male and female
  • Available in PEEK™ and Acetal
  • Machined barb - holds soft wall tubing tight
Dibafit™Dibafit™ Barbed Adapters with 1/4 barbed adaptors are designed for use with soft-wall tubing. Machined from chemically resistant PEEK™ or acetal they have a superior barb with a crisp edge that retains soft wall tubing more securely than molded adaptors. A tight, leak-free connection can be ensured by matching the ID of your tubing to the barb size of the adaptor. If there is not an exact match, always choose the barb size that is just larger than your tubing ID. To connect softwall tubing into any female 1/4”-28 UNF port, use a male ¼”-28 UNF to barb adaptor.
The maleDibafit™ Barbed Adapters with 1/4-28 UNF(F) Connection thread screws into the port and tubing can then be pushed over the barb. To connect softwall tubing to any male 1/4”-28 UNF thread, use a female 1/4”-28 UNF to barb adaptor.The barb is inserted into the softwall tubing and the female ¼ UNF port is available to receive any ¼”-28 UNF threaded fitting. Connecting a male and female adapter allows to create a customzied barbed to barbed adapter.
See Male Connection Barbed Adapter and Female Connection Barbed Adapter options to use with your softwall tubing
Swivel Barbed Adapters Prevent tubing twist during installation and use
  • Free spinning, two-piece adapter
  • Connects to 1/4"-28 flat bottom ports
  • For 1/16", 3/32" and 1/8" ID soft wall tubing
  • Made from Polypropylene
The Upchurch Scientific Swivel Barb AdaptersSwivel Barb Adapters from Upchurch Scientific® are made up of two captive pieces acting as a one-piece fitting for ease of use. Manufactured from Polypropylene and available in three barb sizes, the Swivel Barb will facilitate connection between flexible tubing to a 1/4-28 flat-bottom port. The barbed insert spins freely from the threaded nut in order to prevent the tubing from twisting during installation.