Pure-Fit® Tru Valve System
Pure-Fit® Tru Valve System
Introduction
Pure-Fit® Tru Valve is a multi-function valve system with an innovative design that has your process in mind. The Pure-Fit® Tru Valve system, which is suitable for all flexible tubing materials, provides the user with a solution to efficiency and safety concerns.
Simple to operate and install
The specially designed butterfly mechanism allows installation over tubing and finished assembly. There is no need to take your assembly apart to install the Pure-Fit® Tru Valve. It also offers total flow variability.
Lightweight construction
The Pure-Fit Tru Valve is completely constructed from lightweight borosilicate reinforced nylon material and is also fully autoclavable and sterilizable utilizing gamma irradiation. The lightweight, robust design won’t unbalance your tubing. The superior lightweight construction also allows the fluid floor to be accurately controlled with a calibrated scale on both sides to assist with validation protocols.
Minimizing risk
The Pure-Fit® Tu Valve was not only designed to meter fluid flow, but also as an effective, secure method to close off and lock our a tubing system during a steam-in-place (SIP) cycle. The clamp is to be used with security ties/labels which can be placed and validated as a set on either a closed or preset flow. The ensures that the restricted flow position or secure closure is tamper proof, a significant advantage when ensuring compliance to a Standard Operating Procedure (SOP) is necessary
. Technical data
The Tru Valve Clamp should be placed 1” or less from the face of the molded Tri-Clamp (TC) connection, care should be taken to ensure this is the maximum possible distance. The above tubing sizes were subjected to 100 PSI for 60 minutes at 65 deg F. No failure was seen on multiple testing. These connections can be subjected to SIP at maximum 35 psi. Tru Valve is designed to suit all tubing to .700” OD.