Pure-Fit® TC Tube Clamp System Introduction The Pure-Fit® TC ultimate tube clamp system is the first of a new generation of clamping systems introduced to the pharmaceutical industry. It features a complete outer body with no sharp edges eliminating the potential of snags, scratches or punctures. Unique Design The unique locking mechanism is fully encapsulated to guard against untimely or unwanted release. The Pure-Fit® system has a press down locking system and a side release mechanism allowing for secure and single-handed operation and installation. This high-tech design allows for complete flow stoppage and can be installed over the tube in the normal fashion or assembled after the tube is already in formation. Suited for your needs The Pure-Fit® TC has a high degree of leverage to accommodate varied durometer tubing. It is available in your choice of PVDF, polypropylene, or polysulphone and comes in two standard sizes (3/8” and ¾”) to accommodate a wide range of tubing products. Custom colors and identification are available in this fully autoclavable and sterilizable product. Minimizing risk The locking mechanism is complemented by an integral bore hole which provides total lock-out capability with the use of a cable tie. Pure-Fit® TC meets all USP Class VI criteria and has been physically tested to meet the most demanding applications. |