Ponndorf Hose Pumps
Ponndorf Hose Pumps
What are Ponndorf Hose Pumps?    Ponndorf Hose Pumps in Industrial Applications
Examples of Further Industries Using Ponndorf Hose Pumps
What are Ponndorf Hose Pumps?
Ponndorf Hose Pumps are self-priming rotating positive displacement pumps without any valves. Because of their good suction capacity of up to geodetic suction heights of hs = 8 m they do not need an elevated supply und need not be emptied. The conveying is done in an elastic hose placed along the wall of the pump housing. A rotor is installed centrically in this housing, either with two displacement rollers or shoes arranged at 180¡ or with three of these arranged at 120°.
Pump principle: Ponndorf Hose Pumps of the Series PX and PK
During the suction process the displacement elements are pressing the air resp. the gas from the hose into the pressure tubing and are simultaneouosly sucking the liquid to be pumped into the pump. After the suction process the material is conveyed volumetrically in the hose by the displacement elements from the suction to the pressure side.
Ponndorf hose pumps in industrial applications:
  • Chemical Industry
    Leading companies in the chemical industry are using Ponndorf hose pumps because of their resistance against acids and lyes for the equipment of reliable and economical processing plants. An essential feature for extremely long lifetimes are the chemical-resistant and sturdy pump hoses of Ponndorf.
  • Pharmaceutical and Food Industry
    Ponndorf hose pumps are gaining more and more importance in the food and pharmaceutical industry. Well-known manufacturers of food and pharmaceuticals are meanwhile using Ponndorf pumps for their production plants because of the safe and gentle pumping principle.
  • Another important feature of these pumps is the simple and safe cleaning resp. sterilization by CIP or SIP processes.
  • Waste water Treatment
    As the conveyed material is only in contact with the pump hose, Ponndorf hose pumps are extremely suitable for the pumping of abrasive and solid-containing liquids and sludges. Furthermore, Ponndorf hose pumps are well-proven for feeding oil and fat separation plants because of the gentle and swirl-free conveying of fat/water resp. oil/water mixtures.
  • Construction Industry
    The conveying of concrete, plaster, and cast at construction sites makes great demands on the used pumps. The high quality of the sturdy pump hoses combined with the good suction properties of the Ponndorf hose pumps are the reasons why many well-known construction companies have decided for Ponndorf pumps.
We are always at your disposal for a personal discussion to inform you about further applications of Ponndorf hose pumps.
Examples of further industries using Ponndorf hose pumps:
  • Manufacturing and processing of paints and varnishes
  • Surface refinement, galvanotechnics
  • Wood and paper manufacturing
  • Glass and ceramics industry
You require a reliable, safe and economical solution for your pumping problems? ... we can pump anything able to pass the hose. Give us a challenge!