Hurricanes: The Most Severe Meteorological Phenomenon
Hurricanes:
The Most Severe Meteorological Phenomenon
Hurricane Basics
The ingredients for a hurricane include a pre-existing weather disturbance, warm tropical oceans, moisture, and relatively light winds aloft. If the right conditions persist long enough, they combine to produce violent winds, incredible waves, and torrential rains. Each year, an average of ten tropical storms develop over the Atlantic Ocean, Caribbean Sea, and Gulf of Mexico. Many of these remain over the ocean. Six of these storms become hurricanes each year. In an average 3-year period, roughly five hurricanes strike the United States coastline, killing approximately 50 to 100 people anywhere from Texas to Maine. Of these, two are typically major hurricanes (winds greater than 110 mph).
What is a Hurricane?
Another name for a hurricane is known as the "tropical cyclone." Tropical cyclones are low pressure systems that have thunderstorm activity and rotate counterclockwise. A tropical cyclone that has an organized system of clouds and thunderstorms with a defined surface circulation and maximum sustained winds of 38 mph (33 kt) or less is called a tropical depression. When the tropical cyclone's winds reach 39-73 mph (34-63 kt), it is called a tropical storm. An intense tropical weather system of strong thunderstorms with a well-defined surface circulation and maximum sustained winds of 74 mph (64 kt) or higher is called a hurricane.
The Saffir-Simpson Hurricane Scale defines hurricane strength by categories. A Category 1 storm is the weakest hurricane (winds 74-95 mph or 64-82 kt) while a Category 5 hurricane is the strongest (winds greater than 155 mph or 135 kt). The category of the storm does not necessarily relate directly to the damage it will inflict. Lower category storms (and even tropical storms) can cause substantial damage depending on what other weather features they interact with, where they strike, and how slow they move.
Category Definition–Likely Effects
ONE: Winds 74-95 mph:
No real damage to building structures, Damage primarily to unanchored mobile homes, shrubbery, and trees. Also, some coastal road flooding and minor pier damage.
TWO: Winds 96-110 mph:
Some roofing material, door, and window damage to buildings. Considerable damage to vegetation, mobile homes, and piers. Small craft in unprotected anchorages break moorings.
THREE: Winds 111-130 mph:
Some structural damage to small residences and utility buildings with a minor amount of curtainwall failures, Mobile homes are destroyed. Flooding near the coast destroys smaller structures with larger structures damaged by floating debris. Terrain may be flooded well inland.
FOUR: Winds 131-155 mph:
More extensive curtain wall failures with some complete roof structure failure on small residences. Major erosion of beach areas. Major damage to lower floors of structures near the shore terrain may be flooded well inland.
FIVE: Winds greater than 155 mph:
Complete roof failure on many residences and industrial buildings. Some complete building failures with small utility buildings blown over or away. Major damage to lower floors of all structures located near the shoreline. Massive evacuation of residential areas may be required.
Anatomy of a Hurricane
Hurricanes are normally about 300 miles wide. The eye at a hurricane's center is a relatively calm, clear area approximately 20-40 miles across. The eyewall surrounds the eye, and is composed of dense clouds that contain the highest winds in the storm.
The storm's outer bands (often with hurricane or tropical storm-force winds) are made up of dense thunderstorms ranging from a few miles to tens of miles wide and 50 to 300 miles long.
Hurricane-force winds can extend outward from 25 miles to more than 150 miles. Tropical storm-force winds can stretch out as far as 300 miles from the center of a large hurricane.
Frequently, the right side of a hurricane is the most dangerous in terms of storm surge, winds, and tornadoes. A hurricane's speed and path depend on complex ocean and atmospheric interactions, including the presence or absence of other weather patterns. This complexity of the flow makes it very difficult to predict the speed and direction of a hurricane.
The Birth of a Tropical Cyclone
Tropical cyclones form over warm waters from pre-existing disturbances. These disturbances typically emerge every three or four days from the coast of Africa as "tropical waves" that consist of areas of unsettled weather. Tropical cyclones can also form from the trailing ends of cold fronts and occasionally from upper-level lows. The process by which a tropical cyclone forms and subsequently strengthens into a hurricane depends on at least three conditions:
  1. A pre-existing disturbance with thunderstorms
  2. Warm (at least 80ºF) ocean temperatures to a depth of about 150 feet
  3. Light upper level winds that do not change much in direction and speed throughout the depth of the atmosphere (low wind shear).
Heat and energy for the storm are gathered by the disturbance through contact with warm ocean waters. The winds near the ocean surface spiral into the disturbance's low pressure area. The warm ocean waters add moisture and heat to the air which rises. As the moisture condenses into drops, more heat is released, contributing additional energy to power the storm. Bands of thunderstorms form, and the storm's cloud tops rise higher into the atmosphere. If the winds at these high levels remain relatively light (little or no wind shear), the storm can remain intact and continue to strengthen.
Growth and Maturity
In the early stages, the system appears on the satellite image as a relatively unorganized cluster of thunderstorms. If weather and ocean conditions continue to be favorable, the system can strengthen and become a tropical depression (winds less than 38 mph or 33 kt). At this point, the storm begins to take on the familiar spiral appearance due to the flow of the winds and the rotation of the earth.
If the storm continues to strengthen to tropical storm status (winds 39-73 mph, 34-63 kt), the bands of thunderstorms contribute additional heat and moisture to the storm. The storm becomes a hurricane when winds reach a minimum of 74 mph (64 kt). At this time, the cloud-free hurricane eye typically forms because rapidly sinking air at the center dries and warms the area.
During their life span, hurricanes can last for more than two weeks over the ocean and can travel up the entire Atlantic Coast.
The Storm's End
Just as many factors contribute to the birth of a hurricane, there are many reasons why a hurricane begins to decay. Wind shear can tear the hurricane apart. Moving over cooler water or drier areas can lead to weakening as well. Landfall typically shuts off the hurricane's main moisture source, and the surface circulation can be reduced by friction when it passes over land. Generally, a weakening hurricane or tropical cyclone can reintensify if it moves into a more favorable region or interacts with mid-latitude frontal systems.
Structure
Contrary to how many weather maps appear, a hurricane is more than a point on a weather map, and its path is more than a line. It is a large system that can affect a wide area, requiring that precautions be taken far from where the eye is predicted to come ashore.
The main parts of a hurricane are the rainbands on its outer edges, the eye, and the eyewall. Air spirals in toward the center in a counter-clockwise pattern, and out the top in the opposite direction. In the very center of the storm, air sinks, forming the cloud-free eye.
The hurricane's center, or eye is a relatively calm, clear area usually 20-40 miles across. People in the midst of a hurricane are often amazed at how the incredibly fierce winds and rain can suddenly stop and the sky clear when the eye comes over them. Then, just as quickly, the winds and rain begin again, but this time from the opposite direction.
The dense wall of thunderstorms surrounding the eye has the strongest winds within the storm, this is called the eyewall. Changes in the structure of the eye and eyewall can cause changes in the wind speed, which is an indicator of the storm's intensity. The eye can grow or shrink in size, and double (concentric) eyewalls can form.
The storm's outer rainbands (often with hurricane or tropical storm-force winds) can extend a few hundred miles from the center. Hurricane Andrew's (1992) rainbands reached only 100 miles out from the eye, while those in Hurricane Gilbert (1988) stretched over 500 miles. These dense bands of thunderstorms, which spiral slowly counterclockwise, range in width from a few miles to tens of miles and are 50 to 300 miles long. Sometimes the bands and the eye are obscured by higher level clouds, making it difficult for forecasters to use satellite imagery to monitor the storm.
Hurricane Size
Typical hurricanes are about 300 miles wide although they can vary considerably. Size is not necessarily an indication of hurricane intensity. Hurricane Andrew (1992), the most devastating hurricane of this century, was a relatively small hurricane. Hurricane Danny (left) in 1997 and Hurricane Fran in 1996 show the variability in hurricane size Hurricane destructive winds and rains cover a wide swath. Hurricane-force winds can extend outward to about 25 miles from the storm center of a small hurricane and to more than 150 miles for a large one. The area over which tropical storm-force winds occur is even greater, ranging as far out as almost 300 miles from the eye of a large hurricane.
Hurricane Circulation and Movement
In the northern hemisphere, hurricane winds circulate around the center in a counter-clockwise fashion. This means that the wind direction at your location depends on where the hurricane's eye is. A boat on the northern edge of the orange area in Hurricane Fran would experience winds from the east, while a boat on the southern edge would have westerly winds.
A hurricane's speed and path depend on complex interactions between the storm with its own internal circulations and the earth's atmosphere. The air in which the hurricane is embedded is a constantly moving and changing "river" of air. Other features in that flow, such as high and low pressure systems, can greatly alter the speed and the path of the hurricane. In turn, it can modify the environment around the storm. Typically, a hurricane's forward speed averages around 15-20 mph. However, some hurricanes stall, often causing devastatingly heavy rain. Others can accelerate to more than 60 mph. Hurricane Hazel (1954) hit North Carolina on the morning of 15 October; fourteen hours later it reached Toronto, Canada where it caused 80 deaths. Some hurricanes follow a fairly straight course, while others loop and wobble along the path.
The Right Side of the Storm
As a general rule of thumb, the hurricane's right side (relative to the direction it is travelling) is the most dangerous part of the storm because of the additive effect of the hurricane wind speed and speed of the larger atmospheric flow (the steering winds). The increased winds on the right side increase the storm surge. Tornadoes are also more common here.