How to Read a Hydrometer
How to Read a Hydrometer
  1. Pour your sample into a smooth, clear cylinder or jar that is dry or well rinsed with a portion of the sample.
  2. Make sure your sample is thoroughly mixed before testing.
  3. Immerse the hydrometer in the liquid to a point slightly below the place where it naturally floats. Make sure the hydrometer and liquid are at rest and free of air bubbles.
  4. Measure the temperature of the sample. Ideally, the sample temperature should be equal to the temperature standard of the hydrometer, generally 60°F. If temperature differences are unavoidable, correction tables can help to adjust readings—call our Applications Specialists for information.
  5. Take your reading at the point where the surface of the liquid crosses the hydrometer.
Did You Know...
Degrees Baumé = 145-
145           
sp gr 60°F / 60°F
Degrees API =
141.5         
sp gr 60°F / 60°F
-131.5