How-to Video and Tips for Calibrating your Oakton pH Testr
Video Transcript
Before You Calibrate
Condition your pH tester electrode by immersing it in electrode storage solution, pH 4 buffer, or tap water for at least 30 minutes before use. Do not use deionized water.
Calibration Steps
- Rinse your pH electrode
- Press the on/off button to switch the unit on
- Place the electrode in pH 7 buffer solution
- Press the "Cal" key to put it into calibration mode
The Cal indicator should be shown. The upper display will show the measured reading based on the last calibration. The lower display will indicate the pH standard buffer solution.
- Wait for the reading to stabilize
- Press the "Hold/Ent" key to confirm the first calibration point
If you are doing a single-point calibration, press the "Cal" key at this point to exit calibration mode. If you're doing a multi-point calibration, the lower display will now switch between 4 and 10 the other calibration buffers.
- Rinse the electrode
- Place in pH 4 buffer solution
- Wait for the reading to stabilize
Reading may take up to two minutes to stabilize
- Press the "Hold/Ent" key to confirm the second calibration point
- Place in the final 10 calibration buffer solution
- Wait for the reading to stabilize
- Press the "Hold/Ent" key to confirm the final calibration key
The tester will return to measurement mode. At this point you can rinse the tester and confirm the calibration by placing it in any pH buffer. Or confirm by taking a reading of your sample. Now the calibration is complete. That is how you calibrate your Oakton pH Testr