Emissivity of Specific Materials
Table scrolls horizontally
The emissivity information below was determined under ideal conditions. Note: Surface contamination such as dust, oil films, or other agents will affect the actual emissivity of your material.
Table scrolls horizontally
MATERIAL TEMPERATURE EMISSIVITY °F °C Adobe 68 20 0.90 MATERIAL TEMPERATURE EMISSIVITY °F °C Alloys: Alloy 24ST, polished 75 24 0.90 Alloy 75ST 75 24 0.11 20-Ni, 25-Cr, 55-Fe, oxidized 392 932 200 500 0.90 0.97 60-Ni, 12-Cr, 28-Fe, oxidized 518 1040 270 560 0.89 0.82 80-Ni, 20-Cr, oxidized 212 1112 2372 100 600 1300 0.87 0.87 0.89 MATERIAL TEMPERATURE EMISSIVITY °F °C Aluminum: Unoxidized 77 212 932 25 100 500 0.02 0.03 0.06 Oxidized 390 1110 199 599 0.11 0.19 Oxidized @1110°F 390 1110 199 599 0.11 0.19 Heavily oxidized 200 940 93 504 0.20 0.31 Highly polished 212 100 0.09 Roughly polished 212 100 0.18 Commercial sheet 212 100 0.09 Highly polished plate 440 1070 227 577 0.04 0.06 Bright rolled plate 338 932 170 500 0.04 0.05 Alloy A3003, oxidized 600 900 315 482 0.40 0.40 Alloy 1100-0 200 to 800 93 to 427 0.05 Alloy 24ST 75 24 0.09 Alloy 24ST, polished 75 24 0.09 Alloy 75ST 75 24 0.11 Alloy 75ST, polished 75 24 0.08 MATERIAL TEMPERATURE EMISSIVITY °F °C Asphalt: Pavement 100 38 0.93 Tar paper 68 20 0.93 MATERIAL TEMPERATURE EMISSIVITY °F °C Basalt 68 20 0.72 MATERIAL TEMPERATURE EMISSIVITY °F °C Bismuth: Bright 176 80 0.34 Unoxidized 77 212 25 100 0.05 0.06 MATERIAL TEMPERATURE EMISSIVITY °F °C Brass: Matte 68 20 0.07 Burnished to brown color 68 20 0.40 Cu-Zn, brass oxidized 392 752 1112 200 400 600 0.61 0.60 0.61 Unoxidized 77 25 0.04 Unoxidized 212 100 0.04 MATERIAL TEMPERATURE EMISSIVITY °F °C Brick: Red, rough 70 21 0.93 Gault cream 2500 to 5000 1371 to 2760 0.26 to 0.30 Fire clay 2500 1371 0.75 Light buff 1000 538 0.80 Lime clay 2500 1371 0.43 Fire brick 1832 1000 0.75 to 0.80 Magnesite, refractory 1832 1000 0.38 Gray brick 2012 1100 0.75 Silica, glazed 2000 1093 0.88 Silica, unglazed 2000 1093 0.80 Sandime 2500 to 5000 1371 to 2760 0.59 to 0.63 MATERIAL TEMPERATURE EMISSIVITY °F °C Cadmium 77 25 0.02 MATERIAL TEMPERATURE EMISSIVITY °F °C Carbon: Lamp black 77 25 0.95 Unoxidized 77 25 0.81 Unoxidized 212 932 100 500 0.81 0.79 Candle soot 250 121 0.95 Filament 500 260 0.95 Graphitized 212 572 932 100 300 500 0.76 0.75 0.71 MATERIAL TEMPERATURE EMISSIVITY °F °C Carborundum 1850 1010 0.92 MATERIAL TEMPERATURE EMISSIVITY °F °C Cast iron: Oxidized 390 1110 199 599 0.64 0.78 Unoxidized 212 100 0.21 Strong oxidation 104 482 40 250 0.95 0.95 Liquid 2795 1535 0.29 MATERIAL TEMPERATURE EMISSIVITY °F °C Ceramic: Alumina on Inconel 800 to 2000 427 to 1093 0.69 to 0.45 Earthenware, glazed 70 21 0.90 Earthenware, matte 70 21 0.93 Greens, No. 5210-2C 200 to 750 93 to 399 0.89 to 0.82 Coating, No. C20A 200 to 750 93 to 399 0.73 to 0.67 Porcelain 72 22 0.92 White, Al2O3 200 93 0.90 Zirconia on Inconel 800 to 2000 427 to 1093 0.62 to 0.45 MATERIAL TEMPERATURE EMISSIVITY °F °C Chromium: Chromium 100 1000 38 538 0.08 0.26 Chromium, polished 302 150 0.06 MATERIAL TEMPERATURE EMISSIVITY °F °C Clay: 68 20 0.39 Fired 158 70 0.91 Shale 68 20 0.69 Tiles, light red 2500 to 5000 1371 to 2760 0.32 to 0.34 Tiles, red 2500 to 5000 1371 to 2760 0.40 to 0.51 Tiles, dark purple 2500 to 5000 1371 to 2760 0.78 MATERIAL TEMPERATURE EMISSIVITY °F °C Columbium: Unoxidized 1500 2000 816 1093 0.19 0.24 MATERIAL TEMPERATURE EMISSIVITY °F °C Concrete: Rough 32 to 200 0 to 93 0.94 Tiles, natural 2500 to 5000 1371 to 2760 0.63 to 0.62 Tiles, brown 2500 to 5000 1371 to 2760 0.87 to 0.83 Tiles, black 2500 to 5000 1371 to 2760 0.94 to 0.91 MATERIAL TEMPERATURE EMISSIVITY °F °C Copper: Black, oxidized 100 38 0.78 Etched 100 38 0.09 Matte 100 38 0.22 Roughly polished 100 38 0.07 Polished 100 38 0.03 Highly polished 100 38 0.02 Rolled 100 38 0.64 Rough 100 38 0.74 Molten 1000 1970 2230 538 108 1221 0.15 0.16 0.13 Nickel plated 100 to 500 38 to 260 0.37 MATERIAL TEMPERATURE EMISSIVITY °F °C Cotton cloth 68 20 0.77 MATERIAL TEMPERATURE EMISSIVITY °F °C Dolomite lime 68 20 0.41 MATERIAL TEMPERATURE EMISSIVITY °F °C Dow metal 0 to 600 -18 to 315 0.15 MATERIAL TEMPERATURE EMISSIVITY °F °C Emery corundum 176 80 0.86 MATERIAL TEMPERATURE EMISSIVITY °F °C Glass: Convex D 212 600 932 100 315 500 0.80 0.80 0.76 Nonex 212 600 932 100 315 500 0.82 0.82 0.78 Smooth 32 to 200 0 to 93 0.92 to 0.94 MATERIAL TEMPERATURE EMISSIVITY °F °C Gold: Enamel 212 100 0.37 Plate (0.001) on 0.0005 silver 200 to 750 93 to 399 0.11 to 0.14 Plate (0.001) on 0.005 nickel 200 to 750 93 to 399 0.07 to 0.09 Polished 100 to 500 1000 to 2000 38 to 260 538 to 1093 0.02 0.03 MATERIAL TEMPERATURE EMISSIVITY °F °C Granite 70 21 0.45 MATERIAL TEMPERATURE EMISSIVITY °F °C Gravel 100 38 0.28 MATERIAL TEMPERATURE EMISSIVITY °F °C Gypsum 68 20 0.80 to 0.90 MATERIAL TEMPERATURE EMISSIVITY °F °C Haynes Alloy C., oxidized 600 to 2000 316 to 1093 0.90 to 0.96 MATERIAL TEMPERATURE EMISSIVITY °F °C Haynes Alloy 25, oxidized 600 to 2000 316 to 1093 0.86 to 0.89 MATERIAL TEMPERATURE EMISSIVITY °F °C Haynes Alloy X., oxidized 600 to 2000 316 to 1093 0.85 to 0.88 MATERIAL TEMPERATURE EMISSIVITY °F °C Ice: Smooth 32 0 0.97 Rough 32 0 0.98
https://cmsearch.coleparmer.com/ant_assembler/CPUK/tech_article_landing?format=json&Ntt=/tech-article/emissivity-of-specific-materials&tlg=en-us
go BACK TO TOP