Applying Infrared Thermography to Predictive Maintenance
Applying Infrared Thermography to Predictive Maintenance
Since heat is often an early symptom of equipment damage or malfunction, it makes sense that it is a key performance parameter monitored in predictive maintenance (PdM) programs.
Infrared Thermography Technicians who practice infrared predictive maintenance check the temperature of critical equipment regularly. This allows them to track operating conditions over time and quickly identify unusual readings for further inspection.
By monitoring equipment performance and scheduling maintenance only when needed, facilities reduce the likelihood of unplanned downtime due to equipment failure, spend less on “reactive” maintenance fees and equipment repair costs, and extend the working life of machine assets.
To actually save money, predictive maintenance (PdM) should not create excessive additional maintenance efforts. The goal is to help move maintenance resources away from emergency repairs and into scheduled inspections of key equipment. Inspections take less time than repairs, especially if done with nondestructive testing techniques such as a thermal imager.
Examples of Thermal Imaging Applications
Monitor and measure bearing temperatures in large motors or other rotating equipment
  • Identify “hot spots” in electronic equipment
  • Identify leaks in sealed vessels
  • Find faulty insulation in process pipes or other insulated processes
  • Find faulty terminations in high power electrical circuits
  • Locate overloaded circuit breakers in a power panel
  • Identify fuses at or near their current rated capacity
  • Identify problems in electrical switch gear
A thermal imager takes noncontact infrared (IR) temperature measurements that capture an object’s temperature profile as a two-dimensional picture. Unlike an infrared thermometer that only captures temperature at a single point, a thermal imager can capture temperature from both critical components and the entire integrated unit. Thermal imagers can also store previous and current images for comparison and upload images to a central database.
This article discusses the cost savings of thermal imaging (thermography) PdM, describes how to integrate thermography into a predictive maintenance program, and provides guidelines for successfully capturing and analyzing thermographic data.
Fluke thermal imagers feature IR-Fusion®*, a technology that fuses a visual, or visible light, image with an infrared image for better identification, analysis and image management. The dual images are accurately aligned at any distance heightening details, making it much easier to spot where further investigation is needed.
*The Fluke Ti32 includes SmartView® full analysis and reporting software.
Cost Savings
Studies by the Federal Energy Management Program (FEMP), estimate that a properly functioning predictive maintenance program can provide a savings of 30 to 40 percent over reactive maintenance. Other independent surveys indicate that, on average, starting an industrial predictive maintenance program results in the following savings:
  • Return on investment: 10 times
  • Reduction in maintenance costs: 25 to 30 percent
  • Elimination of breakdowns: 70 to 75 percent
  • Reduction in downtime: 35 to 45 percent
  • Increase in production: 20 to 25 percent
    How to calculate the savings at your facility? Start by estimating the costs of unplanned equipment failures. Factor in personnel resources, costs for parts, and the lost revenue from specific production lines.
Then, once your thermal maintenance program is implemented, you can start to track the savings. Keep records of machine asset availability, production output, and the distribution of maintenance dollars plus total maintenance costs over time. Those numbers will show the return on your thermal imaging and maintenance investment.
Integrating Thermography into PdM
Infrared thermography cameras are the first line of defense in a predictive maintenance program. Technicians can quickly measure and compare heat signatures for each piece of equipment on the inspection route, without disrupting operations.
If the temperature is markedly different from previous readings, facilities can then use other maintenance technologies—vibration testing, motor circuit analysis, airborne ultrasound, and lubricant analysis—to investigate the source of the problem and determine the next course of action.
For best results, integrate all of your maintenance technologies into the same computer system, so that they share the same equipment lists, histories, reports and work orders. Once the infrared data is correlated with data from other technologies, the actual operating condition of all assets can be reported in an integrated format.
Inspection Process
  1. Begin by using existing lists of equipment from a computerized maintenance management system (CMMS) or other inventory tool.
  2. Eliminate items that aren’t well suited for infrared measurement.
  3. Review maintenance and production records. Prioritize key equipment that is prone to failure or often causes production bottlenecks.
  4. Use a database or spreadsheet to group the critical equipment together, either by area or function, into approximately 2- to 3-hour inspection blocks.
  5. Use your thermal imager to capture baseline images of each piece of critical equipment. 
    Note: On some pieces of equipment, you may want to regularly capture multiple thermal images of key components or subsystems.
  6. Download the baseline images into software and document your route with location descriptions, inspection notes, emissivity and RTC levels, and alarm levels if appropriate.
  7. When the next inspection is due, if your thermal imager supports uploading, simply load the previous inspection images onto the camera and follow the onscreen prompts.
This flowchart demonstrates how thermography fits into an overall maintenance program that includes other PdM technologies. (photo courtesy Greg McIntosh, Snell Infrared Canada)
Measurement Guidelines
To capture the best thermal images, follow these best practices:
  • Verify that the target system is operating at a minimum 40 percent of load (lighter loads don’t produce much heat, making it hard to detect problems).
  • Get close to your target and don’t take measurements through doors, especially not through glass. When safety procedures allow, electrical enclosures must be opened or infrared windows or viewports utilized.
  • Account for wind and air currents. These powerful convective forces cool the abnormal hot spots, often below the threshold of detection.
  • Account for ambient air temperatures, especially outdoors. In hot weather, the sun can heat up equipment while cold weather can mask the effects of overheating components.
  • Many problems are hot but not all! Blown fuses and restricted flow in cooling systems are just a couple of examples of situations where a problem is indicated by a cooler than normal signature. In other cases, a cold component is abnormal due to the current being shunted away from the high-resistance connection. Technicians must understand how a machine works and what its heat-related failure signatures are.
  • Consider sources for reflective infrared radiation. Items that have shiny reflective surfaces and are emissive will reflect infrared energy from other nearby objects, including the sun. This can interfere with target temperature measurement and image capture.
  • Unpainted metals are difficult to measure. To improve measurement accuracy and repeatability, consider affixing targets (e.g. paper stickers, electrical tape or painted spots) to such components.
  • Accumulate both numeric temperatures and thermal images to facilitate long-term data analysis. Temperature trends will show you where to investigate more and where inspections can be less frequent.
  • Once you have a database of baseline images, assign an alarm temperature to each one. Upload the most recent version onto your camera before each inspection. If the alarm goes off when you take the new measurement, it indicates a significant change in temperature that needs to be investigated.
Example 1: Spotting Leaky Gaskets and Seals
Finding leaks in sealed vessels is very easy when using a thermal imager. Most leaks develop in or around a gasket or seal. Less often, corrosion will cause a weakness to develop and rupture the vessel.
Either way, an infrared imager can diagnose the problem. To find a leaky gasket or seal, scan along the seal looking for thermal eccentricities. A large change in temperature along the seal or gasket indicates a loss of either heat or cold—the “signature” of a failure.
Example 2: Motor Bearings
Get a baseline image by starting with a newly commissioned and freshly lubricated motor and use your thermal imaging camera to capture an image of the motor bearing housing while the motor is running.
As the motor and its lubrication ages, the bearings wear out and heat-producing friction develops in the motor bearing which will cause the outside of the bearing housing to heat up. Taking additional thermal images at regular intervals and comparing them to the baseline will allow you to analyze the motor’s condition.
When the thermal images indicate an overheating bearing, generate a maintenance order to replace or lubricate the bearing housing; this allows convenient scheduling and prevents unexpected motor failure.
Thermal Measurement Safety
Keeping your thermography inspections accurate, effective and safe is critical. For those reasons it’s also critical that you establish written inspection procedures for measurement collection and interpretation. Following the same steps each time assures you have consistent thermal images in your database for comparison. When creating inspection procedures, refer to the following standards:
National Fire Protection Association (NFPA) 70E safety standard requires that all personnel be educated about the risks they face when working near electrical equipment. Personal protective equipment (PPE) must also be made available to minimize the risk if an accident should occur. For thermographers, PPE generally includes flash­-resistant clothing and a face shield.
The Occupational Safety and Health Administration OSHA 29 CFR, 1910 Subpart S Electrical and Subpart I Personal Protective Equipment Safety standards cover electrical systems, safe work practices, and maintenance requirements.
ISO 6781 International Standards Organization (ISO) (American National Standards Institute) discusses thermal insulation, qualitative detection of thermal irregularities in building envelopes, and infrared methodology.
ASTM International www.astm.org ASTM E 1934, 1213, 1311, 1316, and 1256 Standard guide for examining electrical and mechanical equipment with infrared thermography lists thermography practices and certifications standards. Also reference ASTM 1060 and 1153.
Contact our Application Specialists to assist you with selecting thermal imaging products. Call 1-847-549-7600 or 1-800-323-4340 or go online to ColeParmer.com or chat live.