Published with permission from John Pastor, University of Florida
Aloe vera gel is best known for its therapeutic effect on burned or irritated skin, but in the future you could be eating the gel as a healthful additive to your fruits and veggies. Researchers in Spain say they have developed a gel from the tropical plant that can be used as an edible coating to prolong the quality and safety of fresh produce. The gel, which does not appear to affect food taste or appearance, shows promise as a safe, natural and environmentally-friendly alternative to conventional synthetic preservatives that are currently applied to produce after harvesting, the researchers say.
Although a number of edible coatings have been developed to preserve food freshness, the new coating is believed to be the first to use Aloe vera, according to study leader Daniel Valero, Ph.D., of the University of Miguel Hernández in Alicante, Spain. His study will appear in the Oct. 5 print issue of the American Chemical Society’s Journal of Agricultural and Food Chemistry, the Society’s peer-reviewed publication.
Valero and his associates dipped a group of common table grapes (Crimson Seedless) into Aloe vera gel and stored them for five weeks under low temperature while exposing a group of untreated table grapes to the same conditions. The colorless Aloe gel used in this study was developed through a special processing technique that maximized the amount of active compounds in the gel, Valero and associates say. The gel can also be applied as a spray, they add.
The untreated grapes appeared to deteriorate rapidly within about 7 days, whereas the gel-coated grapes were well-preserved for up to 35 days under the same experimental conditions, the researchers say. The gel-treated grapes were firmer, had less weight loss and less color change than the untreated grapes, measures which correspond to higher freshness, they say.
A sensory panel (10 people) evaluated the quality of both the untreated and the gel-treated grapes by consuming some of the grapes. They found that the gel-treated grapes were generally superior in taste.
The researchers believe that the gel works through a combination of mechanisms. Composed mostly of polysaccharides, the gel appears to act as a natural barrier to moisture and oxygen, which can speed food deterioration. But the gel also enhances food safety, the scientists say. Based on previous studies by others, Aloe vera gel appears to contain various antibiotic and antifungal compounds that can potentially delay or inhibit microorganisms that are responsible for foodborne illness in humans as well as food spoilage.
Although the health effects of Aloe gel on human consumption were not directly measured in this study, the coating is believed to be safe, the researchers say. They note that Aloe vera gel has been used as a functional ingredient in some foods and beverages for years. In addition to preserving table grapes, which are highly perishable, the gel can be applied to other fruits and vegetables, they say. Further testing of the gel on other types of produce is anticipated. The gel also offers potential environmental benefits, the researchers add. It could provide a greener alternative to sulfur dioxide and other synthetic food preservatives that are commonly used on produce and increasingly the target of health concerns, they say. Valero and his associates have filed a patent application in Spain for their gel. It could appear in the U.S. consumer market within a year, Valero estimates, noting that the gel is a natural product and is unlikely to face any major regulatory hurdles. Funding for this study was provided by the Spanish Ministry of Science and Technology and the European Commission via FEDER (European fund for regional development).
The American Chemical Society is a nonprofit organization, chartered by the U.S. Congress, with a multidisciplinary membership of more than 158,000 chemists and chemical engineers. It publishes numerous scientific journals and databases, convenes major research conferences and provides educational, science policy and career programs in chemistry. Its main offices are in Washington, D.C., and Columbus, Ohio. Mark T. Sampson