Wiretrol® II Glass Capillary Micropipettes
Micropipettes provide accuracy and convenience with dual graduations
  • Dual graduations for noncontact delivery
  • ±1% accuracy
Draw fluid into the calibrated glass barrel with fingertip plunger. Accuracy is ±1% of contained volume. Each pack contains 250 disposable glass barrels and two plungers: use long plunger to clear barrel; use short plunger to maintain air cushion between plunger and sample.
£69.61 - £89.81GBP / Pkg of 250
£83.53 - £107.77GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Wetted Matterial
Availability Pricing
Wiretrol II Capillary Micropipettes, Glass, 1-5 µL; 250/Pk 0795100 Mfr # 5-000-2005
COMPARE
5 1 Glass
£89.81GBP / Pkg of 250
£107.77GBP (incl VAT)
Wiretrol II Capillary Micropipettes, Glass, 5-10 µL; 250/Pk 0795105 Mfr # 5-000-2010
COMPARE
10 5 Glass
£69.61GBP / Pkg of 250
£83.53GBP (incl VAT)
Drummond 5-000-2200 Positive Displacement Microdispensers, 100-200 µ, 250/pk 0795125 Mfr # 5-000-2200
COMPARE
200 100