Wilmad-LabGlass Standard 5 mm NMR Tube Caps
Easily track samples with different colored caps
  • Durable ethylene vinyl acetate construction
  • Compatible with 5 mm OD NMR tubes
These Wilmad-LabGlass standard 5 mm NMR tube caps come in a package of assorted colors. The different colors help track samples based on various reference solvents. These caps are compatible with 5 mm OD NMR tubes and are available in either an assortment pack of 100 or 1000 caps. When using chloroform or acetone as a reference solution, avoid direct contact between the sample and cap and long-term storage over two hours.
£8.87 - £56.83GBP / Pkg of 1000
£10.64 - £68.20GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cap Style Packaging Description
Availability Pricing
SP Wilmad Caps for Disposable NMR Tubes, Assorted Color; 100/Pk 8399962 Mfr # 521-ASST-100
COMPARE
Disposable for 5 mm OD NMR tubes 100/Pk Caps for Disposable NMR Tubes, Assorted Color; 100/Pk
£8.87GBP / Pkg of 100
£10.64GBP (incl VAT)
SP Wilmad Caps for Disposable NMR Tubes, Assorted Color; 1000/Pk 8399988 Mfr # 521-ASST-1000
COMPARE
Disposable for 5 mm OD NMR tubes 1000/Pk Caps for Disposable NMR Tubes, Assorted Color; 1000/Pk
£56.83GBP / Pkg of 1000
£68.20GBP (incl VAT)