Wilmad-LabGlass High-Throughput NMR Sample Tubes with Caps
High-throughput NMR tubes have one of the best OD tolerances in the industry
  • NMR tubes are constructed of ASTM E438 Type 1 Class B borosilicate glass
  • Ensure spectrum clarity with an averaged camber of 60 micron
These Wilmad-LabGlass high-throughput NMR tubes have an averaged camber of 60 micron to guarantee the spectrum resolution for small molecules (MW<250) samples up to 600 MHz. Each 5mm tube meets precise wall thickness to ensure they fit tightly in spinner turbine. All tubes are made from borosilicate glass. Choose from tubes with authentic Bruker® SampleJet® caps without code or tubes with disposable caps.
£137.15 - £285.82GBP / Pkg of 100
£164.58 - £342.98GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Tube Length (mm) Tube Outside Diameter (OD) (mm)
Availability Pricing
Wilmad-LabGlass High-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L; 100/Pk WZ-83999-58 Mfr # WG-1000-7
COMPARE
Disposable 178 4.94665
£137.15GBP / Pkg of 100
£164.58GBP (incl VAT)
Wilmad-LabGlass High-Throughput NMR Tube and Cap, 4.94665 mm OD x 8" L; 100/Pk WZ-83999-60 Mfr # WG-1000-8
COMPARE
Disposable 230.2 4.94665
£164.69GBP / Pkg of 100
£197.63GBP (incl VAT)
Wilmad-LabGlass High-Throughput NMR Tube and Cap, 4.94665 mm OD x 7" L, Large Marking Spot; 100/Pk WZ-83999-61 Mfr # WG-1003-7
COMPARE
Disposable 178 4.94665
£137.15GBP / Pkg of 100
£164.58GBP (incl VAT)
Wilmad-LabGlass Bruker® SampleJet® High-Throughput NMR Tube and Cap, 5 mm OD x 103.5 mm L; 100/Pk WZ-83999-63 Mfr # WG-1000-4-SJ
COMPARE
Bruker® SampleJet® 103.5 5
£162.35GBP / Pkg of 100
£194.82GBP (incl VAT)
Wilmad-LabGlass Bruker® SampleJet® High-Throughput NMR Tube and Cap, 5 mm OD x 178 mm L; 100/Pk WZ-83999-90 Mfr # WG-1000-7-SJ
COMPARE
Bruker® SampleJet® 178 5
£285.82GBP / Pkg of 100
£342.98GBP (incl VAT)