Watson-Marlow 120U Variable-Speed Pump System
Compact pump systems with remote control features deliver accuracy and repeatability to optimize your fluid handling process
  • Remotely control start/stop, speed, and pump direction
  • Small, stackable pumps require minimal benchtop space
  • Control and view speed on digital LCD
  • Keypad lock prevents setting accidental changes
  • Smooth IP31-rated housing for easy wipedown
  • Auto key for changing between manual and auto control
Easily maintain fluid integrity using peristaltic pumps. Only the bore of the tubing contacts the fluid, eliminating the risk of the pump contaminating the fluid or the fluid contaminating the pump. Peristaltic pumps are great for use with most liquids, including viscous, shear-sensitive, corrosive, and abrasive fluids.
Manually or remotely control these compact pumps for accurate and repeatable fluid transfer. Each pump system includes a single-channel pump head to deliver flow rates from 0.001 to 170 mL/min*—select from 2-roller, 4-roller, or 4-roller flip-top models. Simple manual control allows start/stop, direction change, and speed adjustment from the keypad; adjust speed in 0.1 rpm increments. Press the MAX key for rapid priming; AUTO START for automatic power recovery. Remotely control start/stop, pump direction, and speed using analog signals (4 to 20 mA, 0 to 10 V); 0 to 5 V output allows remote speed display.
Use the keypad lock feature to prevent any unwanted changes to pump settings. A smooth, IP31-rated housing encases a maintenance-free, brushless motor for quick and easy cleaning. Contact us for free technical application support on pump & tubing selection.
£3,217.74 - £3,867.70GBP / Each
£3,861.29 - £4,641.24GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Pump Head
Availability Pricing
Watson-Marlow 120U/D1 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Single-Channel, 4-Roller; 90 to 264 VAC 5013005 Mfr # 010.6141.010
COMPARE
120 0.001 400D1 four-roller
£3,217.74GBP / Each
£3,861.29GBP (incl VAT)
Watson-Marlow 120U/DV Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Flip-Top Head, Single-Channel, 4-Roller; 90 to 264 VAC 5013006 Mfr # 010.6141.DA0
COMPARE
170 0.002 114DV four-roller, flip-top
£3,263.51GBP / Each
£3,916.21GBP (incl VAT)
Watson-Marlow 120U/R Variable-Speed Pump System, Manual/Analog Control, 0.1 to 32 rpm, Single-Channel, 2-Roller; 90 to 264 VAC 5013009 Mfr # 010.6141.R00
COMPARE
54 0.003 102R two-roller
£3,226.90GBP / Each
£3,872.28GBP (incl VAT)
Watson-Marlow 120U/DM3 Variable-Speed Pump System, Manual/Analog Control, 0.1 to 200 rpm, Three-Channel, 4-Roller; 90 to 264 VAC 5014200 Mfr # 010.6141.M30
COMPARE
36 0.0001 400DM3 four-roller
£3,867.70GBP / Each
£4,641.24GBP (incl VAT)