Versilon® PTFE Tubing
High-temperature PTFE tubing for chemically inert, high-performance fluid handling
  • Exceptional temperature resistance up to ~260°C (500°F) supports demanding thermal applications
  • Chemically inert PTFE construction resists aggressive media across a wide range of industrial and laboratory uses
  • Smooth, dimensionally stable tubing ensures consistent flow and reliable performance
  • Suitable for diverse applications including chemical transfer, gas delivery, and industrial fluid handling
Versilon® PTFE tubing is designed for applications requiring extreme temperature resistance and broad chemical compatibility. Manufactured from fully fluorinated polytetrafluoroethylene, this tubing offers excellent inertness, making it ideal for handling aggressive chemicals, gases, and solvents across a variety of industries. With a maximum operating temperature of up to approximately 260°C (500°F) and stability at very low temperatures, it performs reliably in both high-heat and cryogenic environments. Its smooth, dimensionally stable construction supports consistent fluid transfer, while its versatility makes it suitable for applications ranging from chemical processing and gas delivery to automotive and electrical systems.
£83.65 - £1,604.55GBP / Pkg of 1
£100.38 - £1,925.46GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Versilon PTFE Tubing, 1/16" ID x 1/8" OD, 50 ft 4852624 Mfr # TSPTFE-0125-031-50
COMPARE
1/16 1/8 1/32
£83.65GBP / Pkg of 1
£100.38GBP (incl VAT)
Versilon PTFE Tubing, 1/8" ID x 3/16" OD, 50 ft 4852625 Mfr # TSPTFE-0187-031-50
COMPARE
1/8 3/16 1/16
£110.90GBP / Pkg of 1
£133.08GBP (incl VAT)
Versilon PTFE Tubing, 3/16" ID x 1/4" OD, 50 ft 4852626 Mfr # TSPTFE-0250-031-50
COMPARE
3/16 1/4 1/32
£159.10GBP / Pkg of 1
£190.92GBP (incl VAT)
Versilon PTFE Tubing, 5/32" ID x 1/4" OD, 50 ft 4852627 Mfr # TSPTFE-0250-047-50
COMPARE
5/32 1/4 3/64
£263.65GBP / Pkg of 1
£316.38GBP (incl VAT)
Versilon PTFE Tubing, 1/8" ID x 1/4" OD, 50 ft 4852628 Mfr # TSPTFE-0250-062-50
COMPARE
1/8 1/4 1/16
£289.10GBP / Pkg of 1
£346.92GBP (incl VAT)
Versilon PTFE Tubing, 1/4" ID x 5/16" OD, 50 ft 4852629 Mfr # TSPTFE-0312-031-50
COMPARE
1/4 5/16 1/32
£263.65GBP / Pkg of 1
£316.38GBP (incl VAT)
Versilon PTFE Tubing, 3/16" ID x 5/16" OD, 50 ft 4852630 Mfr # TSPTFE-0312-062-50
COMPARE
3/16 5/16 1/16
£418.20GBP / Pkg of 1
£501.84GBP (incl VAT)
Versilon PTFE Tubing, 5/16" ID x 3/8" OD, 50 ft 4852631 Mfr # TSPTFE-0375-031-50
COMPARE
5/16 3/8 1/32
£268.20GBP / Pkg of 1
£321.84GBP (incl VAT)
Versilon PTFE Tubing, 1/4" ID x 3/8" OD, 50 ft 4852632 Mfr # TSPTFE-0375-062-50
COMPARE
1/4 3/8 1/16
£586.35GBP / Pkg of 1
£703.62GBP (incl VAT)
Versilon PTFE Tubing, 3/8" ID x 7/16" OD, 50 ft 4852633 Mfr # TSPTFE-0437-031-50
COMPARE
3/8 7/16 1/16
£327.25GBP / Pkg of 1
£392.70GBP (incl VAT)
Versilon PTFE Tubing, 5/16" ID x 7/16" OD, 50 ft 4852634 Mfr # TSPTFE-0437-062-50
COMPARE
5/16 7/16 1/16
£836.35GBP / Pkg of 1
£1,003.62GBP (incl VAT)
Versilon PTFE Tubing, 7/16" ID x 1/2" OD, 50 ft 4852635 Mfr # TSPTFE-0500-031-50
COMPARE
7/16 1/2 1/32
£368.20GBP / Pkg of 1
£441.84GBP (incl VAT)
Versilon PTFE Tubing, 3/8" ID x 1/2" OD, 50 ft 4852636 Mfr # TSPTFE-0500-062-50
COMPARE
3/8 1/2 1/16
£845.45GBP / Pkg of 1
£1,014.54GBP (incl VAT)
Versilon PTFE Tubing, 1/2" ID x 5/8" OD, 50 ft 4852637 Mfr # TSPTFE-0625-062-50
COMPARE
1/2 5/8 1/16
£1,186.35GBP / Pkg of 1
£1,423.62GBP (incl VAT)
Versilon PTFE Tubing, 5/8" ID x 3/4" OD, 50 ft 4852638 Mfr # TSPTFE-0750-062-50
COMPARE
5/8 3/4 1/16
£1,604.55GBP / Pkg of 1
£1,925.46GBP (incl VAT)
Showing 1- 15 of 15