Vanguard Safety Uncoated Cut Resistant Work Gloves
Extended cuff protects wrists and forearms
  • Seamless knit HPPE blended liner keeps hands cool and dry
  • Ambidextrous
  • FFDCA compliant for food handling
  • EN 388 Standard with 3X41D rating
  • ANSI abrasion level 3
  • ANSI A4 cut level
  • Machine washable
These A4 uncoated work gloves provide excellent dexterity for use in the food processing, food service, and manufacturing environments. An extended cuff protects wrists and forearms. The gloves may be wore as a cut protection liner under disposable and chemical resistant gloves.
£140.00GBP / Case of 48
£168.00GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Lining Size
Availability Pricing
Vanguard Safety R18G410 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, XL; 24 Pairs/CS 7890716 Mfr # R18G410
COMPARE
10 15/64 13g High Performance Polyethylene (HPPE) Extra large
£140.00GBP / Case of 48
£168.00GBP (incl VAT)
Vanguard Safety R18G47 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, Small; 24 Pairs/CS 7890717 Mfr # R18G47
COMPARE
9 13g High Performance Polyethylene (HPPE) Small
£140.00GBP / Case of 48
£168.00GBP (incl VAT)
Vanguard Safety R18G48 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, Medium; 24 Pairs/CS 7890718 Mfr # R18G48
COMPARE
9 29/64 13g High Performance Polyethylene (HPPE) Medium
£140.00GBP / Case of 48
£168.00GBP (incl VAT)
Vanguard Safety R18G49 A4 HPPE Uncoated Cut Resistant Work Gloves, Gray, Large; 24 Pairs/CS 7890719 Mfr # R18G49
COMPARE
9 27/32 13g High Performance Polyethylene (HPPE) Large
£140.00GBP / Case of 48
£168.00GBP (incl VAT)