Value Plastics Tee Tube Fitting with 200 Series Barb
A secure connection under pressure with 200 series barbs
  • Quick and easy assembly and disassembly without the need for tools
  • Barbed ridges provide secure grip on the inside of flexible tubing
  • Tee orientation provides flexibility and versatility
The Value Plastics 200 Series Tee Hose Barb Fittings are designed to provide outstanding performance characteristics when used with flexible tubing. Tubing is expanded by 50% above its nominal inside diameter, permitting high-pressure capabilities. The tee orientation offers directional flexibility, while the barbs grip the inside of the hose, creating a tight seal to prevent leaks or disconnection. These adapters are available in various materials to accommodate various applications and compliance requirements.
£35.97 - £55.92GBP / Pkg of 25
£43.16 - £67.10GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
Value Plastics Hose Barb Fitting, Tee Union, PVDF, 200 Series Barb, 1/8" ID; 25/Pk 5012088 Mfr # T230-J2A
COMPARE
1/8 PVDF Hose Barb Fitting, Tee Union, PVDF, 200 Series Barb, 1/8" ID; 25/Pk
£55.92GBP / Pkg of 25
£67.10GBP (incl VAT)
Value Plastics Hose Barb Fitting, Tee Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk 5012094 Mfr # T230-6005
COMPARE
1/8 Polypropylene Hose Barb Fitting, Tee Union, Polypropylene, 200 Series Barb, 1/8" ID; 25/Pk
£35.97GBP / Pkg of 25
£43.16GBP (incl VAT)