Value Plastics Sanitary Fitting Caps, Polypropylene
Seal sanitary ports with a secure and leak-proof connection
  • Ideal for capping off fluid transfer system
  • Sanitary fittings provide a secure connection where preventing contamination is crucial
  • Constructed of ADCF polypropylene
These Value Plastics Sanitary Fitting Caps are designed to easily seal sanitary ports. Sanitary fittings ensure a reliable connection while facilitating easy cleaning and preventing the buildup of bacteria or other contaminants. Sanitary end caps are often used in food and beverage, pharmaceutical, and biotechnology applications where maintaining cleanliness is crucial. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
£1,115.56GBP / Pkg of 100
£1,338.67GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connection Type Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
Value Plastics Sanitary Fitting, Straight Adapter, Polypropylene, 3/4" Mini Sanitary x Cap; 100/Pk 5011867 Mfr # SFMNC-6005
COMPARE
Cap 0.75
£1,115.56GBP / Pkg of 100
£1,338.67GBP (incl VAT)
Value Plastics Sanitary Fitting, Straight Adapter, Polypropylene, 1-1/2" Sanitary x Cap; 100/Pk 5011872 Mfr # SFMXC-6005
COMPARE
Cap 1.5
£1,115.56GBP / Pkg of 100
£1,338.67GBP (incl VAT)