Value Plastics Male Luer with Integral Lock Ring to 500 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The integral lock ring is a built-in mechanism that provides a secure connection to the female luer to prevent accidental disconnection and stop leakage or contamination. The 500 series barb is composed of blended radii giving it a distinct look. Each barb is designed to accommodate a range of tube sizes and tube types. Consideration should be given to flow restriction, pressure capability, and the force required for assembly of the tube. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
£15.65 - £25.47GBP / Pkg of 25
£18.78 - £30.56GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/16" ID; 25/Pk 5012015 Mfr # MTLL004-1
COMPARE
1/16 Nylon
£15.65GBP / Pkg of 25
£18.78GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 500 Series Barb 1/16" ID; 25/Pk 5012016 Mfr # MTLL004-6005
COMPARE
1/16 Polypropylene
£21.52GBP / Pkg of 25
£25.82GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/8" ID; 25/Pk 5012017 Mfr # MTLL013-1
COMPARE
1/8 Nylon
£17.07GBP / Pkg of 25
£20.48GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 500 Series Barb 1/8" ID; 25/Pk 5012018 Mfr # MTLL013-6005
COMPARE
1/8 Polypropylene
£22.32GBP / Pkg of 25
£26.78GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer Integral Lock Ring x 500 Series Barb 1/4" ID; 25/Pk 5012019 Mfr # MTLL055-1
COMPARE
1/4 Nylon
£19.43GBP / Pkg of 25
£23.32GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring x 500 Series Barb 1/4" ID; 25/Pk 5012020 Mfr # MTLL055-6005
COMPARE
1/4 Polypropylene
£25.47GBP / Pkg of 25
£30.56GBP (incl VAT)