Value Plastics Male Luer with Integral Lock Ring to 400 Series Barb
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
  • Rotating locks minimize twisting and kinking of tubing during positioning
  • Tapered cone seals are reliable and uncomplicated
  • Facilitate single-use configurations that minimize downtime
These luer fittings come in many configurations and several materials, joining tubing from 1/16” (1.6 mm) ID to 1/4” (6.4 mm) ID. The integral lock ring is a built-in mechanism that provides a secure connection to the female luer to prevent accidental disconnection and stop leakage or contamination. The 400 series barb provides easy assembly while maintaining excellent pressure and tensile capabilities. Originally created for semi-rigid tubing, this series is preferred for applications where tensile/pressure properties are not highly critical, or when ease of assembly is desired. The 400 series expands the tubing 25% above its nominal inside diameter. Because the fittings are made to ISO standards that are well understood globally, it ensures proper interconnections and often removes the need to retrofit*.
Options include nylon, polycarbonate, or polypropylene resin.
Common features of nylon include a high mechanical strength, great balance of stiffness and toughness, good performance at high temperatures, good electrical and flammability properties, and resistance against abrasion or chemicals. It can be sterilized using gamma radiation, Ethylene oxide (EtO), or autoclave.
Polypropylene is clarified and produced without any animal derived components or phthalates. It can be sterilized using gamma radiation, Ethylene oxide (EtO), E-beam, or autoclave.
Polycarbonate is transparent and features high impact, chemical, UV, and electrical resistance. Sterilization is only recommended via ethylene oxide (EtO). Gamma radiation up to 10 Mrads is possible with minor loss of physical properties – it will discolor to a yellow-green hue. Autoclave is not recommended and may crack the fitting.
£17.87 - £23.47GBP / Pkg of 25
£21.44 - £28.16GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Integral Lock Ring x 400 Series Barb 1/16" ID; 25/Pk 5012034 Mfr # MTLL410-9
COMPARE
1/16 Polycarbonate
£21.16GBP / Pkg of 25
£25.39GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, White Nylon, Male Luer with Integral Lock Ring x 400 Series Barb 3/32" ID; 25/Pk 5012072 Mfr # MTLL420-1
COMPARE
3/32 Nylon
£17.87GBP / Pkg of 25
£21.44GBP (incl VAT)
Value Plastics Luer to Hose Barb Fitting, Straight Adapter, Polypropylene, Male Luer with Integral Lock Ring x 400 Series Barb 3/32" ID; 25/Pk 5012073 Mfr # MTLL420-6005
COMPARE
3/32 Polypropylene
£23.47GBP / Pkg of 25
£28.16GBP (incl VAT)