Value Plastics Male Luer Slip to Female Luer Thread Adapter Fitting
Luer fittings allow for a simple yet secure connection
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
The Male Luer Slip to Female Luer Thread Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
£22.84 - £25.99GBP / Pkg of 25
£27.41 - £31.19GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
Value Plastics Luer Fitting, Straight Adapter, Polypropylene, Male Luer Slip x Female Luer Thread; 25/Pk 5012005 Mfr # LC78-6005
COMPARE
Polypropylene Male Luer Slip Female Luer Thread
£25.99GBP / Pkg of 25
£31.19GBP (incl VAT)
Value Plastics Luer Fitting, Straight Adapter, Clear Polycarbonate, Male Luer Slip x Female Luer Thread; 25/Pk 5012006 Mfr # LC78-9
COMPARE
Polycarbonate Male Luer Slip Female Luer Thread
£22.84GBP / Pkg of 25
£27.41GBP (incl VAT)