Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Coupler
Prevent leaks with integral lock ring mechanism
  • Luer fittings allow for a simple yet secure twist-lock connection
  • Easily transition from a male luer to female luer fitting
  • Ideal for general laboratory and single-use applications
Value Plastics Male Luer Integral Lock Ring to Female Luer Thread Style Couplers are designed to easily transition your fluid transfer system from a male to female luer connection. Luer fittings ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The integral lock ring is a feature built into the male luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. These adapters are available in various materials to accommodate various applications and compliance requirements.
£21.79 - £27.57GBP / Pkg of 25
£26.15 - £33.08GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
Value Plastics Luer Fitting, White Nylon, Male Luer Integral Lock Ring x Female Luer Thread; 25/Pk 5012003 Mfr # LC34-1
COMPARE
Nylon Male Luer Integral Lock Ring Female Luer Thread
£21.79GBP / Pkg of 25
£26.15GBP (incl VAT)
Value Plastics Luer Fitting, Straight Adapter, Polypropylene, Male Luer Integral Lock Ring to Female Luer Thread; 25/Pk 5012004 Mfr # LC34-6005
COMPARE
Polypropylene Male Luer Integral Lock Ring Female Luer Thread
£27.57GBP / Pkg of 25
£33.08GBP (incl VAT)