Value Plastics Luer Adapter Fittings, Tee, Polypropylene
Easily transition to a luer fitting with directional flexibility
  • Quick and easy assembly and disassembly without the need for tools
  • Luer fittings allow for a simple yet secure connection
  • Conforms to ISO-594-1 and ISO 594-2 standards
  • Constructed of ADCF polypropylene
These Value Plastics Luer Elbow Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The tee orientation offers directional flexibility, creating a tight seal to prevent leaks or disconnection. Transparent polypropylene (PP) is suitable for Gamma radiation, ethylene oxide (EtO), E-beam, and autoclave sterilization.
May be used with separate rotating lock ring, 50115-54, sold separately.
£63.53GBP / Pkg of 25
£76.24GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Male Luer Slip x Male Luer Slip x Female Luer Thread; 25/Pk 5012010 Mfr # LT787-6005
COMPARE
Female Luer Thread Male Slip Luer
£63.53GBP / Pkg of 25
£76.24GBP (incl VAT)
Value Plastics Luer Fitting, Tee Adapter, Polypropylene, Female Luer Thread x Female Luer Thread x Male Luer Slip; 25/Pk 5012011 Mfr # LT878-6005
COMPARE
Female Luer Thread Female Luer Thread
£63.53GBP / Pkg of 25
£76.24GBP (incl VAT)