Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Luer fittings allow for a simple yet secure connection
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
The Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. Adapters are available in various materials to accommodate various applications and compliance requirements.
£28.88 - £40.17GBP / Pkg of 25
£34.66 - £48.20GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Luer Connection Type (Port 1) Luer Connection Type (Port 2)
Availability Pricing
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Female Luer Thread Style with 1/4" Hex x 10-32 UNF(M); 25/Pk 5012000 Mfr # KFTLL-1
COMPARE
Nylon Female Luer Thread Style with 1/4" Hex 10-32 UNF(M) Thread
£28.88GBP / Pkg of 25
£34.66GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 1/4" Hex x 10-32 UNF(M); 25/Pk 5012001 Mfr # KFTLL-9
COMPARE
Polycarbonate Female Luer Thread Style with 1/4" Hex 10-32 UNF(M) Thread
£40.17GBP / Pkg of 25
£48.20GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk 5012035 Mfr # BSFTLL-1
COMPARE
Nylon Female Luer Thread Style with 5/16" Hex 1/4-28 UNF(M) Bottom Sealing Thread
£30.19GBP / Pkg of 25
£36.23GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk 5012084 Mfr # SFTLL-1
COMPARE
Nylon Female Luer Thread Style with 5/16" Hex 1/4-28 UNF(M) Thread
£28.88GBP / Pkg of 25
£34.66GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk 5012085 Mfr # SFTLL-6005
COMPARE
Polypropylene Female Luer Thread Style with 5/16" Hex 1/4-28 UNF(M) Thread
£38.49GBP / Pkg of 25
£46.19GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, Clear Polycarbonate, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M); 25/Pk 5012086 Mfr # SFTLL-9
COMPARE
Polycarbonate Female Luer Thread Style with 5/16" Hex 1/4-28 UNF(M) Thread
£40.17GBP / Pkg of 25
£48.20GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 5/16" Hex x 1/4-28 UNF(M) Bottom Sealing Thread; 25/Pk 5012092 Mfr # BSFTLL-6005
COMPARE
Polypropylene Female Luer Thread Style with 5/16" Hex 1/4-28 UNF(M) Bottom Sealing Thread
£39.38GBP / Pkg of 25
£47.26GBP (incl VAT)
Showing 1- 7 of 7