Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Luer fittings allow for a simple yet secure connection
  • Quick and easy assembly and disassembly without the need for tools
  • Thread design provides a secure closure—preventing contamination
  • Conforms to ISO-594-1 and ISO 594-2 standards
The Female Luer Thread Style to NPT Adapter Fittings are designed to ensure a tight and reliable connection while allowing for quick and easy assembly and disassembly without the need for tools. The thread feature is built into the female luer fitting that ensures the fittings remain firmly connected until intentionally disconnected. NPT (National Pipe Thread) is a type of tapered thread commonly used for sealing and connecting pipe fittings and components. NPT thread fittings are designed to create a tight seal when assembled, preventing leakage of fluids or gases under pressure. Adapters are available in various materials to accommodate various applications and compliance requirements.
£42.27 - £55.65GBP / Pkg of 25
£50.72 - £66.78GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Luer Connection Type (Port 2) Regulatory Approvals Thread Size NPT(M) (Port 2) (InchShort)
Availability Pricing
Value Plastics Luer to Threaded Fitting, Straight Adapter, White Nylon, Female Luer Thread  7/16" Hex x 1/8-27 NPT(M); 25/Pk 5011994 Mfr # 18FTLL-1
COMPARE
1/8-27 NPT(M) Thread Non-DEHP 1/8
£42.27GBP / Pkg of 25
£50.72GBP (incl VAT)
Value Plastics Luer to Threaded Fitting, Straight Adapter, Polypropylene, Female Luer Thread Style with 7/16" Hex x 1/8-27 NPT(M); 25/Pk 5011995 Mfr # 18FTLL-6005
COMPARE
1/8-27 NPT(M) Thread FDA 1/8
£55.65GBP / Pkg of 25
£66.78GBP (incl VAT)