Tygon® SPT-3350 Platinum-Cured Silicone Tubing
Ultra-smooth inner surface reduces particle entrapment
  • Ideal for food and beverage transfer
  • Minimal extractable maintains fluid integrity
  • Up to three times smoother than other silicone tubing
  • FDA 21 CFR 175.300 compliant
  • Free technical application support available to assist with tubing selection
Tygon® SPT-3350 sanitary platinum-cured silicone tubing features a platinum curing process designed to meet the demanding sanitary standards of food and beverage industry. To improve the flow of fluids, the inner surface of the tube is designed to reduce the risk of particle entrapment and microscopic build-up during fluid transfer. The smooth inner surface aids in complete cleaning and sterilization.
The tubing is made from 100% synthetic virgin silicone. No plant or animal products or by-products are used in the manufacture of, nor intentionally added to the tubing. They are produced in an ISO 14644 Class 7 clean room facility from USP 88 Class VI and FDA 21CFR approved raw materials in accordance with cGMP principles.
£86.28 - £3,545.54GBP / Pkg of 1
£103.54 - £4,254.65GBP (incl VAT)
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/32" ID x 3/32" OD; 50 Ft WZ-50113-89 Mfr # ABW01NSF
COMPARE
1/32 3/32 1/32
£86.28GBP / Pkg of 1
£103.54GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/16" ID x 1/8" OD; 50 Ft WZ-50113-90 Mfr # ABW02NSF
COMPARE
1/16 1/8 1/32
£97.53GBP / Pkg of 1
£117.04GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/16" ID x 3/16" OD; 50 Ft WZ-50113-91 Mfr # ABW03NSF
COMPARE
1/16 3/16 1/16
£132.69GBP / Pkg of 1
£159.23GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/32" ID x 5/32" OD; 50 Ft WZ-50113-92 Mfr # ABW04NSF
COMPARE
3/32 5/32 1/32
£105.50GBP / Pkg of 1
£126.60GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/32" ID x 7/32" OD; 50 Ft WZ-50113-93 Mfr # ABW05NSF
COMPARE
3/32 7/32 1/16
£156.14GBP / Pkg of 1
£187.37GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/8" ID x 3/16" OD; 50 Ft WZ-50113-94 Mfr # ABW06NSF
COMPARE
1/8 3/16 1/32
£113.47GBP / Pkg of 1
£136.16GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft WZ-50113-95 Mfr # ABW07NSF
COMPARE
1/8 1/4 1/16
£165.04GBP / Pkg of 1
£198.05GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 5/32" ID x 7/32" OD; 50 Ft WZ-50113-96 Mfr # ABW09NSF
COMPARE
5/32 7/32 1/32
£119.10GBP / Pkg of 1
£142.92GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/16" ID x 1/4" OD; 50 Ft WZ-50113-97 Mfr # ABW11NSF
COMPARE
3/16 1/4 1/32
£140.66GBP / Pkg of 1
£168.79GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft WZ-50113-98 Mfr # ABW12NSF
COMPARE
3/16 5/16 1/16
£210.99GBP / Pkg of 1
£253.19GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft WZ-50113-99 Mfr # ABW13NSF
COMPARE
3/16 3/8 3/32
£301.95GBP / Pkg of 1
£362.34GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/16" ID x 7/16" OD; 50 Ft WZ-50114-00 Mfr # ABW14NSF
COMPARE
3/16 7/16 1/8
£416.35GBP / Pkg of 1
£499.62GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/4" ID x 5/16" OD; 50 Ft WZ-50114-01 Mfr # ABW16NSF
COMPARE
1/4 5/16 1/32
£161.29GBP / Pkg of 1
£193.55GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft WZ-50114-02 Mfr # ABW17NSF
COMPARE
1/4 3/8 1/16
£231.62GBP / Pkg of 1
£277.94GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft WZ-50114-03 Mfr # ABW18NSF
COMPARE
1/4 7/16 3/32
£353.53GBP / Pkg of 1
£424.24GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft WZ-50114-04 Mfr # ABW19NSF
COMPARE
1/4 1/2 1/8
£478.24GBP / Pkg of 1
£573.89GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 5/16" ID x 7/16" OD; 50 Ft WZ-50114-05 Mfr # ABW22NSF
COMPARE
5/16 7/16 1/16
£287.89GBP / Pkg of 1
£345.47GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 5/16" ID x 1/2" OD; 50 Ft WZ-50114-06 Mfr # ABW23NSF
COMPARE
5/16 1/2 3/32
£406.98GBP / Pkg of 1
£488.38GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft WZ-50114-07 Mfr # ABW27NSF
COMPARE
3/8 1/2 1/16
£323.52GBP / Pkg of 1
£388.22GBP (incl VAT)
Tygon SPT-3350 Platinum-Cured Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft WZ-50114-08 Mfr # ABW28NSF
COMPARE
3/8 9/16 3/32
£460.43GBP / Pkg of 1
£552.52GBP (incl VAT)
Showing 1 - 20 of 32