Tygon® 3370 I.B. Braid-Reinforced Platinum-Cured Silicone Tubing
Designed to transfer high-purity fluids under pressure
  • Braid reinforcement handles high pressures
  • Platinum cured silicone—shows exceptional biocompatibility
  • Low extractables—perfect for maintaining fluid purity
  • Smooth inner surface minimizes particle entrapment
  • Exhibits excellent flexibility, durability, and chemical and temperature resistance
  • Withstands repeated CIP and SIP
  • USP Class VI, 3A Sanitary Standard, and FDA 21 CFR Part 177.2600 compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
Tygon 3370 I.B. silicone tubing, produced from a platinum curing process, meets the most demanding requirements of biocompatibility. It features a smooth, nonporous inner surface that promotes easy filling, draining, and transferring of fluids without trapping any particles that can cause bacterial growth. Braid reinforcement offers increased pressure resistance. With a low extractable content, it is an ideal choice for transferring high-purity fluids under pressure.
£1,031.12 - £3,408.89GBP / Pkg of 1
£1,237.34 - £4,090.67GBP (incl VAT)
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 3/4" ID x 1-5/32" OD; 50 Ft 5020120 Mfr # AHJ001671
COMPARE
3/4 1 5/32 1/5
£2,591.12GBP / Pkg of 1
£3,109.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 3/16" ID x 7/16" OD; 50 Ft 5020121 Mfr # AHJ001718
COMPARE
3/16 7/16 1/8
£1,031.12GBP / Pkg of 1
£1,237.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 5020122 Mfr # AHJ001719
COMPARE
1/4 33/64 1/8
£1,191.12GBP / Pkg of 1
£1,429.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 3/8" ID x 11/16" OD; 50 Ft 5020123 Mfr # AHJ001720
COMPARE
3/8 11/16 5/32
£1,391.12GBP / Pkg of 1
£1,669.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 1/2" ID x 27/32" OD; 50 Ft 5020124 Mfr # AHJ001721
COMPARE
1/2 27/32 11/64
£1,884.45GBP / Pkg of 1
£2,261.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 5/8" ID x 31/32" OD; 50 Ft 5020125 Mfr # AHJ001722
COMPARE
5/8 63/64 11/64
£2,271.12GBP / Pkg of 1
£2,725.34GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 1" ID x 1-25/64" OD; 50 Ft 5020126 Mfr # AHJ001724
COMPARE
1 1 25/64 3/16
£3,408.89GBP / Pkg of 1
£4,090.67GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 1-1/4" ID x 1-21/32" OD; 25 Ft 5020127 Mfr # AHJ421725
COMPARE
1 1/4 1 41/64 3/16
£1,955.56GBP / Pkg of 1
£2,346.67GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 1-1/2" ID x 1-29/32" OD; 25 Ft 5020128 Mfr # AHJ421726
COMPARE
1 1/2 1 29/32 1/5
£2,382.23GBP / Pkg of 1
£2,858.68GBP (incl VAT)
Tygon 3370 I.B. Reinforced Platinum-Cured Silicone Tubing, 2" ID x 2-7/16" OD; 25 Ft 5020129 Mfr # AHJ421798
COMPARE
2 2 7/16 7/32
£2,577.78GBP / Pkg of 1
£3,093.34GBP (incl VAT)
Showing 1- 10 of 10