Tygon® 3350 Platinum-Cured Sanitary Silicone Tubing
Ultra-smooth inner surface—perfect for sanitary transfer of sensitive fluids
  • Smooth inner surface minimizes particle entrapment
  • Platinum cured silicone—shows exceptional biocompatibility
  • Low extractables—perfect for maintaining fluid purity
  • Excellent fluid flow characteristics due to reduced surface area
  • USP Class VI, 3A Sanitary Standard, FDA 21 CFR Part 177.2600, and NSF 51 compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection cleaning and sterilization
Designed with ultra-smooth inner bore, the Tygon 3350 platinum-cured silicone tubing is perfect for sanitary transfer of sensitive fluids. The smooth inner surface ensures minimal particle entrapment and microscopic build-up during fluid transfer while low extractables help maintain fluid integrity. The smooth bore also aids in fluid flow by reducing the surface area.
£89.78 - £3,551.12GBP / Pkg of 1
£107.74 - £4,261.34GBP (incl VAT)
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/32" ID x 3/32" OD; 50 Ft 5020088 Mfr # ABW00001
COMPARE
1/32 3/32 1/32
£89.78GBP / Pkg of 1
£107.74GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/16" ID x 1/8" OD; 50 Ft 5020089 Mfr # ABW00002
COMPARE
1/16 1/8 1/32
£109.78GBP / Pkg of 1
£131.74GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/16" ID x 3/16" OD; 50 Ft 5020090 Mfr # ABW00003
COMPARE
1/16 3/16 1/16
£150.15GBP / Pkg of 1
£180.18GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/32" ID x 5/32" OD; 50 Ft 5020091 Mfr # ABW00004
COMPARE
3/32 5/32 1/32
£118.45GBP / Pkg of 1
£142.14GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/32" ID x 7/32" OD; 50 Ft 5020092 Mfr # ABW00005
COMPARE
3/32 7/32 1/16
£163.34GBP / Pkg of 1
£196.01GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/8" ID x 3/16" OD; 50 Ft 5020093 Mfr # ABW00006
COMPARE
1/8 3/16 1/32
£123.34GBP / Pkg of 1
£148.01GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft 5020094 Mfr # ABW00007
COMPARE
1/8 1/4 1/16
£172.67GBP / Pkg of 1
£207.20GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 5/32" ID x 7/32" OD; 50 Ft 5020095 Mfr # ABW00009
COMPARE
5/32 7/32 1/32
£126.00GBP / Pkg of 1
£151.20GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/16" ID x 1/4" OD; 50 Ft 5020096 Mfr # ABW00011
COMPARE
3/16 1/4 1/32
£149.34GBP / Pkg of 1
£179.21GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft 5020097 Mfr # ABW00012
COMPARE
3/16 5/16 1/16
£236.25GBP / Pkg of 1
£283.50GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft 5020098 Mfr # ABW00013
COMPARE
3/16 3/8 3/32
£312.89GBP / Pkg of 1
£375.47GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/16" ID x 7/16" OD; 50 Ft 5020099 Mfr # ABW00014
COMPARE
3/16 7/16 1/8
£429.34GBP / Pkg of 1
£515.21GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/4" ID x 5/16" OD; 50 Ft 5020100 Mfr # ABW00016
COMPARE
1/4 5/16 1/32
£170.00GBP / Pkg of 1
£204.00GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft 5020101 Mfr # ABW00017
COMPARE
1/4 3/8 1/16
£259.35GBP / Pkg of 1
£311.22GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft 5020102 Mfr # ABW00018
COMPARE
1/4 7/16 3/32
£364.00GBP / Pkg of 1
£436.80GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 5020103 Mfr # ABW00019
COMPARE
1/4 1/2 1/8
£490.67GBP / Pkg of 1
£588.80GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 5/16" ID x 7/16" OD; 50 Ft 5020104 Mfr # ABW00022
COMPARE
5/16 7/16 1/16
£318.67GBP / Pkg of 1
£382.40GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 5/16" ID x 1/2" OD; 50 Ft 5020105 Mfr # ABW00023
COMPARE
5/16 1/2 3/32
£420.00GBP / Pkg of 1
£504.00GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft 5020106 Mfr # ABW00027
COMPARE
3/8 1/2 1/16
£336.00GBP / Pkg of 1
£403.20GBP (incl VAT)
Tygon 3350 Platinum-Cured Sanitary Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft 5020107 Mfr # ABW00028
COMPARE
3/8 9/16 3/32
£472.00GBP / Pkg of 1
£566.40GBP (incl VAT)
Showing 1 - 20 of 32