TSI VelociCalc Air Velocity Meters
Thermoanemometers with fast and accurate airflow measurements
  • Displays up to three measurements simultaneously
  • Available with manual or continuous data logging
  • Logs up to 12,700 samples and 100 test IDs
These meters simultaneously measure and datalog several ventilation parameters using a single probe with multiple sensors. Great for cleanroom filtration airflow measurements — they automatically calculate volumetric flow rate for round, square, and rectangular areas. All models measure velocity, temperature and calculate flow.
£1,298.38 - £1,974.52GBP / Each
£1,558.06 - £2,369.42GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
TSI 9535 VelociCalc Air Velocity Meter with Straight Probe, no Humidity Sensor 1027125 Mfr # 9535
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
£1,298.38GBP / Each
£1,558.06GBP (incl VAT)
TSI 9545 VelociCalc Air Velocity Meter with Straight Probe and Humidity Sensor 1027127 Mfr # 9545
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
£1,738.08GBP / Each
£2,085.70GBP (incl VAT)
TSI 9535-A VelociCalc Air Velocity Meter; Articulating Probe/Manual Log 1027135 Mfr # 9535-A
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
£1,514.08GBP / Each
£1,816.90GBP (incl VAT)
TSI 9545-A VelociCalc Air Velocity Meter; Articulating Probe/Auto Log 1027137 Mfr # 9545-A
COMPARE
±3% of reading or ±3 ft/min (0.05 m/s) (whichever is greater) 1 ft/min (0.01 m/s) 6000
£1,974.52GBP / Each
£2,369.42GBP (incl VAT)