Traceable® Vane Thermoanemometers with Calibration
Rugged, compact meter with ergonomic-grip design
  • Easy-to-use keypad
  • O-ring seal and membrane keypad keep out dust and dirt
  • Recalls min/max readings
  • Press HOLD key to freeze current reading
Compact vane thermoanemometers are designed for intuitive, instruction-free operation. Press the HOLD button to freeze display reading for easy recording of results. Set to Record Mode to capture minimum and maximum readings. The tough ABS plastic housing with O-ring seal and membrane keypad keeps dust and dirt out, allowing long service life even in harsh environments. All models measure airflow and temperature and feature a high contrast LCD display with 3/8”-high digits.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
£143.00 - £328.38GBP / Each
£171.60 - £394.06GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Measurement Parameter
Availability Pricing
Traceable Vane Thermoanemometer with Calibration 3795512 Mfr # 3650TR
COMPARE
Temperature and air velocity
£143.00GBP / Each
£171.60GBP (incl VAT)
Traceable Vane Thermoanemometer/Humidity/Dew Point with Calibration 3795514 Mfr # 3651TR
COMPARE
Temperature, air velocity, humidity, and dew point
£168.26GBP / Each
£201.91GBP (incl VAT)
Traceable Vane Thermoanemometer/Humidity/Dew Point/Barometric Pressure with Calibration 3795516 Mfr # 3652TR
COMPARE
Temperature, air velocity, humidity,dew point, and barometric pressure
£328.38GBP / Each
£394.06GBP (incl VAT)