Traceable® Mini-Alarm Timer/Stopwatch
Program to count up or down—to 100 hours!
  • Features: count up/down, time in/out, memory, extra-loud alarm, clock
  • Numbered keys make it a breeze to set countdown time
  • When zero is reached, alarm sounds TIMES UP flashes on display and timer counts up time elapsed since alarm
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Count up/down timer can be programmed to count up or down to 100 hours with a 1-second interval. After the count down cycle, the timer counts up to indicate elapsed time and sounds an audible alarm when time has elapsed. Memory recalls the last setting for easy reuse. Timer can double as an alarm clock.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
£40.13GBP / Each
£48.16GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Resolution Timing Capacity
Availability Pricing
Traceable Mini-Alarm Timer/Stopwatch 9446004 Mfr # 5021TR
COMPARE
±8.64 sec/day 1 sec 100 hours
£40.13GBP / Each
£48.16GBP (incl VAT)