Traceable® Digital Pocket Thermometers with Calibration
Get an updated reading every 10 seconds
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Wide temperature range from -58 to 572°F (-50 to 300°C)
  • Large, bright LCD with 3/8"H digits
  • 4-3/4"L x 0.19" dia stainless steel stem fits nicely into included protective sleeve
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
£32.25 - £65.35GBP / Each
£38.70 - £78.42GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
572 -58 Calibrated Digital Pocket Thermometer Ultra; ±0.4°C Accuracy at Tested Points
£65.35GBP / Each
£78.42GBP (incl VAT)
COMPARE
572 -58 Digital Pocket Thermometer with Calibration; ±1°C accuracy
£32.25GBP / Each
£38.70GBP (incl VAT)