Thermo Scientific Orion™ Single Junction pH/ATC Combination Electrodes
Accurately measure pH and temperature together in just one electrode
  • Convenient built-in temperature sensor takes temperature measurements
  • Epoxy body for ruggedness
Use these silver chloride electrodes to accurately measure pH of samples. Electrodes also feature a built-in ATC for temperature measurements.
£172.32 - £456.05GBP / Each
£206.78 - £547.26GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Reference Junction
Availability Pricing
Thermo Scientific 9157BN Triode™ 3-in-1 Combination pH/ATC Electrode, Epoxy Body, Glass Fiber, Refillable, BNC 5881990 Mfr # 9157BN
COMPARE
90 Glass fiber
£456.05GBP / Each
£547.26GBP (incl VAT)
Thermo Scientific 9147BN Combination pH/ATC Electrode, Epoxy Body, HDPE Pin, (Requires BNC to USS Adapter) 5882292 Mfr # 9147BN
COMPARE
50 HDPE pin
£172.32GBP / Each
£206.78GBP (incl VAT)