Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
High-purity, pharma-grade tubing designed for transferring fluids in pharmaceutical, laboratory, and bioprocess applications
  • Exhibits very good resilience and elastic memory
  • Odorless, noncytotoxic, and nonhemolytic
  • Low-binding surface helps minimize particle and odor absorption
  • Cleanroom packed and double bagged package
  • Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants
  • Meets USP <661> Physiochemical test
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Use this pharma-grade platinum-cured silicone tubing for general fluid transfer and peristaltic pumping applications in pharmaceutical, laboratory, and bioprocess manufacturing. A low-binding surface of the tubing minimizes flavor, odor, and protein absorption, helping to maintain the integrity of the fluid. Excellent elastic memory and resiliency coupled with low hysteresis and compression set ensure that fluid transfers are reliable and accurate every time.
£106.05 - £1,117.40GBP / Pkg of 1
£127.26 - £1,340.88GBP (incl VAT)
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/16" ID x 1/8" OD; 50 Ft 0013841 Mfr # 8600-0020
COMPARE
1/16 1/8 1/32
£106.05GBP / Pkg of 1
£127.26GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/8" ID x 1/4" OD; 50 Ft 0013842 Mfr # 8600-0030
COMPARE
1/8 1/4 1/16
£171.20GBP / Pkg of 1
£205.44GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/16" ID x 5/16" OD; 50 Ft 0013843 Mfr # 8600-0040
COMPARE
3/16 5/16 1/16
£215.90GBP / Pkg of 1
£259.08GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/16" ID x 3/8" OD; 50 Ft 0013844 Mfr # 8600-0050
COMPARE
3/16 3/8 3/32
£323.50GBP / Pkg of 1
£388.20GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/4" ID x 3/8" OD; 50 Ft 0013845 Mfr # 8600-0060
COMPARE
1/4 3/8 1/16
£247.75GBP / Pkg of 1
£297.30GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/4" ID x 7/16" OD; 50 Ft 0013846 Mfr # 8600-0070
COMPARE
1/4 7/16 3/32
£398.50GBP / Pkg of 1
£478.20GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/4" ID x 1/2" OD; 50 Ft 0013847 Mfr # 8600-0080
COMPARE
1/4 1/2 1/8
£511.35GBP / Pkg of 1
£613.62GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 5/16" ID x 1/2" OD; 50 Ft 0013848 Mfr # 8600-0100
COMPARE
5/16 1/2 3/32
£435.60GBP / Pkg of 1
£522.72GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/8" ID x 1/2" OD; 50 Ft 0013849 Mfr # 8600-0120
COMPARE
3/8 1/2 1/16
£330.30GBP / Pkg of 1
£396.36GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/8" ID x 9/16" OD; 50 Ft 0013850 Mfr # 8600-0130
COMPARE
3/8 9/16 3/32
£492.40GBP / Pkg of 1
£590.88GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/8" ID x 5/8" OD; 50 Ft 0013851 Mfr # 8600-0140
COMPARE
3/8 5/8 1/8
£640.15GBP / Pkg of 1
£768.18GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/2" ID x 11/16" OD; 50 Ft 0013852 Mfr # 8600-0190
COMPARE
1/2 11/16 3/32
£723.50GBP / Pkg of 1
£868.20GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 1/2" ID x 3/4" OD; 50 Ft 0013853 Mfr # 8600-0200
COMPARE
1/2 3/4 1/8
£1,000.00GBP / Pkg of 1
£1,200.00GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 5/8" ID x 7/8" OD; 50 Ft 0013854 Mfr # 8600-0260
COMPARE
5/8 7/8 1/8
£958.35GBP / Pkg of 1
£1,150.02GBP (incl VAT)
Thermo Scientific Nalgene Pharma-Grade Platinum-Cured Silicone Tubing, 3/4" ID x 1" OD; 50 Ft 0013855 Mfr # 8600-0310
COMPARE
3/4 1 1/8
£1,117.40GBP / Pkg of 1
£1,340.88GBP (incl VAT)
Showing 1- 15 of 15