Thermo Scientific™ Nalgene™ Metric DEHP-Free PVC Tubing
Made from high-quality resins and plasticizers, othophthalate-free PVC tubing offers exceptional purity
  • Exhibits excellent chemical resistance—resists hardening and discoloration from a wide range of laboratory chemicals
  • Does not pass on odor or taste
  • Clear tubing—view fluid being transferred
  • 60 Shore A durometer
  • Complies with FDA CFR21 for food packaging and USP VI
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made from high-quality resins and plasticizers, the DEHP-free, clear PVC tubing is ideal for use in a wide range of pharmaceutical, research, and general laboratory applications. Tubing resists hardening and discoloration while exhibiting dimensional stability when exposed to various laboratory chemicals. Tubing ID and regulatory compliance marked at every 10 cm interval make for easy tubing identification.
£299.63 - £748.15GBP / Pkg of 1
£359.56 - £897.78GBP (incl VAT)
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (mm) Tubing OD (mm) Wall Thickness (mm)
Availability Pricing
Thermo Scientific Nalgene 8703-0407 Metric DEHP-Free PVC Tubing, 4.0 mm ID x 7.0 mm OD; 25 m 5014166 Mfr # 8703-0407
COMPARE
4 7 1.5
£299.63GBP / Pkg of 1
£359.56GBP (incl VAT)
Thermo Scientific Nalgene 8703-0508 Metric DEHP-Free PVC Tubing, 5.0 mm ID x 8.0 mm OD; 25 m 5014167 Mfr # 8703-0508
COMPARE
5 8 1.5
£397.04GBP / Pkg of 1
£476.45GBP (incl VAT)
Thermo Scientific Nalgene 8703-0609 Metric DEHP-Free PVC Tubing, 6.0 mm ID x 9.0 mm OD; 25 m 5014168 Mfr # 8703-0609
COMPARE
6 9 1.5
£528.15GBP / Pkg of 1
£633.78GBP (incl VAT)
Thermo Scientific Nalgene 8703-0610 Metric DEHP-Free PVC Tubing, 6.0 mm ID x 10.0 mm OD; 25 m 5014169 Mfr # 8703-0610
COMPARE
6 10 2
£593.34GBP / Pkg of 1
£712.01GBP (incl VAT)
Thermo Scientific Nalgene 8703-0710 Metric DEHP-Free PVC Tubing, 7.0 mm ID x 10.0 mm OD; 25 m 5014170 Mfr # 8703-0710
COMPARE
7 10 1.5
£643.71GBP / Pkg of 1
£772.45GBP (incl VAT)
Thermo Scientific Nalgene 8703-0811 Metric DEHP-Free PVC Tubing, 8.0 mm ID x 11.0 mm OD; 25 m 5014171 Mfr # 8703-0811
COMPARE
8 11 1.5
£678.52GBP / Pkg of 1
£814.22GBP (incl VAT)
Thermo Scientific Nalgene 8703-1520 Metric DEHP-Free PVC Tubing, 15.0 mm ID x 20.0 mm OD; 10 m 5014176 Mfr # 8703-1520
COMPARE
15 20 2.5
£420.00GBP / Pkg of 1
£504.00GBP (incl VAT)
Thermo Scientific Nalgene 8703-0812 Metric DEHP-Free PVC Tubing, 8.0 mm ID x 12.0 mm OD; 25 m 5014182 Mfr # 8703-0812
COMPARE
8 12 2
£685.93GBP / Pkg of 1
£823.12GBP (incl VAT)
Thermo Scientific Nalgene 8703-0913 Metric DEHP-Free PVC Tubing, 9.0 mm ID x 13.0 mm OD; 25 m 5014183 Mfr # 8703-0913
COMPARE
9 13 2
£748.15GBP / Pkg of 1
£897.78GBP (incl VAT)
Showing 1- 9 of 9