Thermo Scientific™ Nalgene™ DEHP-Free PVC Vacuum Tubing
Thick-walled tubing can withstand a full vacuum of 760 mm Hg (29.9" Hg) at room temperature
  • Othophthalate-free PVC tubing offers exceptional purity
  • Exhibits excellent chemical resistance—resists hardening and discoloration from a wide range of laboratory chemicals
  • Does not pass on odor or taste
  • Clear tubing—view fluid being transferred
  • 60 Shore A durometer
  • Complies with FDA CFR21 for food packaging and USP VI
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made from high-quality resins and plasticizers, the DEHP-free, clear PVC tubing is ideal for use in a wide range of pharmaceutical, research, and general laboratory applications. Tubing resists hardening and discoloration while exhibiting dimensional stability when exposed to various laboratory chemicals. Tubing ID and regulatory compliance marked at every 12" interval make for easy tubing identification.
£105.00 - £385.19GBP / Pkg of 1
£126.00 - £462.23GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Thermo Scientific Nalgene 8702-0055 DEHP-Free PVC Vacuum Tubing, 3/16" ID x 9/16" OD; 50 Ft 5014162 Mfr # 8702-0055
COMPARE
3/16 9/16 3/16
£385.19GBP / Pkg of 1
£462.23GBP (incl VAT)
Thermo Scientific Nalgene 8702-1065 DEHP-Free PVC Vacuum Tubing, 1/4" ID x 5/8" OD; 10 Ft 5014163 Mfr # 8702-1065
COMPARE
1/4 5/8 3/16
£105.00GBP / Pkg of 1
£126.00GBP (incl VAT)
Thermo Scientific Nalgene 8702-1145 DEHP-Free PVC Vacuum Tubing, 3/8" ID x 7/8" OD; 10 Ft 5014164 Mfr # 8702-1145
COMPARE
3/8 7/8 1/4
£186.67GBP / Pkg of 1
£224.00GBP (incl VAT)
Thermo Scientific Nalgene 8702-1210 DEHP-Free PVC Vacuum Tubing, 1/2" ID x 1-1/8" OD; 10 Ft 5014165 Mfr # 8702-1210
COMPARE
1/2 1 1/8 5/16
£268.52GBP / Pkg of 1
£322.22GBP (incl VAT)