Thermo Scientific™ Nalgene™ DEHP-Free PVC Tubing
Made from high-quality resins and plasticizers, othophthalate-free PVC tubing offers exceptional purity
  • Exhibits excellent chemical resistance—resists hardening and discoloration from a wide range of laboratory chemicals
  • Does not pass on odor or taste
  • Clear tubing—view fluid being transferred
  • 60 Shore A durometer
  • Complies with FDA CFR21 for food packaging and USP VI
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made from high-quality resins and plasticizers, the DEHP-free, clear PVC tubing is ideal for use in a wide range of pharmaceutical, research, and general laboratory applications. Tubing resists hardening and discoloration while exhibiting dimensional stability when exposed to various laboratory chemicals. Tubing ID and regulatory compliance marked at every 12" interval (except 50141-46) make for easy tubing identification.
£36.97 - £597.04GBP / Pkg of 1
£44.36 - £716.45GBP (incl VAT)
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
Thermo Scientific Nalgene 8701-0020 DEHP-Free PVC Tubing, 1/8" ID x 1/4" OD; 50 Ft 2205073 Mfr # 8701-0020
COMPARE
1/8 1/4 1/16
£72.75GBP / Pkg of 1
£87.30GBP (incl VAT)
Thermo Scientific Nalgene 8701-0030 DEHP-Free PVC Tubing, 3/16" ID x 5/16" OD; 50 Ft 2205074 Mfr # 8701-0030
COMPARE
3/16 5/16 1/16
£106.12GBP / Pkg of 1
£127.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0010 DEHP-Free PVC Tubing, 1/8" ID x 3/16" OD; 50 Ft 5014145 Mfr # 8701-0010
COMPARE
1/8 3/16 1/32
£36.97GBP / Pkg of 1
£44.36GBP (incl VAT)
Thermo Scientific Nalgene 8701-0060 DEHP-Free PVC Tubing, 1/4" ID x 3/8" OD; 50 Ft 5014146 Mfr # 8701-0060
COMPARE
1/4 3/8 1/16
£124.45GBP / Pkg of 1
£149.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0070 DEHP-Free PVC Tubing, 1/4" ID x 7/16" OD; 50 Ft 5014147 Mfr # 8701-0070
COMPARE
1/4 7/16 3/32
£194.45GBP / Pkg of 1
£233.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0080 DEHP-Free PVC Tubing, 1/4" ID x 1/2" OD; 50 Ft 5014148 Mfr # 8701-0080
COMPARE
1/4 1/2 1/8
£237.41GBP / Pkg of 1
£284.89GBP (incl VAT)
Thermo Scientific Nalgene 8701-0090 DEHP-Free PVC Tubing, 5/16" ID x 7/16" OD; 50 Ft 5014149 Mfr # 8701-0090
COMPARE
5/16 7/16 1/16
£140.00GBP / Pkg of 1
£168.00GBP (incl VAT)
Thermo Scientific Nalgene 8701-0100 DEHP-Free PVC Tubing, 5/16" ID x 1/2" OD; 50 Ft 5014150 Mfr # 8701-0100
COMPARE
5/16 1/2 3/32
£206.12GBP / Pkg of 1
£247.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0110 DEHP-Free PVC Tubing, 5/16" ID x 9/16" OD; 50 Ft 5014151 Mfr # 8701-0110
COMPARE
5/16 9/16 1/8
£248.89GBP / Pkg of 1
£298.67GBP (incl VAT)
Thermo Scientific Nalgene 8701-0120 DEHP-Free PVC Tubing, 3/8" ID x 1/2" OD; 50 Ft 5014152 Mfr # 8701-0120
COMPARE
3/8 1/2 1/16
£159.45GBP / Pkg of 1
£191.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0130 DEHP-Free PVC Tubing, 3/8" ID x 9/16" OD; 50 Ft 5014153 Mfr # 8701-0130
COMPARE
3/8 9/16 3/32
£210.00GBP / Pkg of 1
£252.00GBP (incl VAT)
Thermo Scientific Nalgene 8701-0140 DEHP-Free PVC Tubing, 3/8" ID x 5/8" OD; 50 Ft 5014154 Mfr # 8701-0140
COMPARE
3/8 5/8 1/8
£303.34GBP / Pkg of 1
£364.01GBP (incl VAT)
Thermo Scientific Nalgene 8701-0160 DEHP-Free PVC Tubing, 7/16" ID x 5/8" OD; 50 Ft 5014155 Mfr # 8701-0160
COMPARE
7/16 5/8 3/32
£245.19GBP / Pkg of 1
£294.23GBP (incl VAT)
Thermo Scientific Nalgene 8701-0180 DEHP-Free PVC Tubing, 1/2" ID x 5/8" OD; 50 Ft 5014156 Mfr # 8701-0180
COMPARE
1/2 5/8 1/16
£198.34GBP / Pkg of 1
£238.01GBP (incl VAT)
Thermo Scientific Nalgene 8701-0190 DEHP-Free PVC Tubing, 1/2" ID x 11/16" OD; 50 Ft 5014157 Mfr # 8701-0190
COMPARE
1/2 11/16 3/32
£284.08GBP / Pkg of 1
£340.90GBP (incl VAT)
Thermo Scientific Nalgene 8701-0200 DEHP-Free PVC Tubing, 1/2" ID x 3/4" OD; 50 Ft 5014158 Mfr # 8701-0200
COMPARE
1/2 3/4 1/8
£377.78GBP / Pkg of 1
£453.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0260 DEHP-Free PVC Tubing, 5/8" ID X 7/8" OD; 50 Ft 5014159 Mfr # 8701-0260
COMPARE
5/8 7/8 1/8
£420.00GBP / Pkg of 1
£504.00GBP (incl VAT)
Thermo Scientific Nalgene 8701-0310 DEHP-Free PVC Tubing, 3/4" ID x 1" OD; 50 Ft 5014160 Mfr # 8701-0310
COMPARE
3/4 1 1/8
£477.78GBP / Pkg of 1
£573.34GBP (incl VAT)
Thermo Scientific Nalgene 8701-0350 DEHP-Free PVC Tubing, 1" ID x 1-1/4" OD; 50 Ft 5014161 Mfr # 8701-0350
COMPARE
1 1 1/4 1/8
£597.04GBP / Pkg of 1
£716.45GBP (incl VAT)
Showing 1- 19 of 19